Skip to Content
Merck
All Photos(1)

Key Documents

F0778

Sigma-Aldrich

Felbamate

Synonym(s):

2-Phenyl-1,3-propanediol dicarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O4
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

solubility

alcohol: soluble

SMILES string

NC(=O)OCC(COC(N)=O)c1ccccc1

InChI

1S/C11H14N2O4/c12-10(14)16-6-9(7-17-11(13)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,12,14)(H2,13,15)

InChI key

WKGXYQFOCVYPAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anticonvulsant agent that is an allosteric antagonist at the NR2B subunit of the NMDA glutamate receptor; also has γ-aminobutyric acid (GABAA) receptor agonist properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Brown et al.
Critical reviews in neurobiology, 12(3), 205-222 (1998-12-10)
Felbamate was launched in 1993 in the U.S. as a "new generation" antiepileptic drug (AED) with a unique mechanism of action. It proved efficacious in patients refractory to other AEDs and was particularly beneficial in children suffering from Lennox-Gastaut syndrome
K D Laxer
The Western journal of medicine, 161(3), 309-314 (1994-09-01)
For the first time in 15 years, new antiepileptic medications are available for the treatment of patients with seizure disorders. These drugs have demonstrated efficacy in animal models of epilepsy and in controlled clinical trials. Felbamate was licensed in 1993
Huai-Ren Chang et al.
Biophysical journal, 93(6), 1981-1992 (2007-05-22)
Felbamate (FBM) is a potent nonsedative anticonvulsant whose clinical effect is chiefly related to gating modification (and thus use-dependent inhibition) rather than pore block of N-methyl-D-aspartate (NMDA) channels at pH 7.4. Using whole-cell recording in rat hippocampal neurons, we examined
John M Pellock et al.
Epilepsy research, 71(2-3), 89-101 (2006-08-08)
An expert panel convened to evaluate data and review current clinical practices regarding the novel antiepileptic drug (AED) felbamate. Felbamate has demonstrated efficacy against a variety of refractory seizures types, including seizures associated with Lennox-Gastaut syndrome, but postmarketing experience revealed
Angelique M Leone et al.
Chemical research in toxicology, 20(4), 600-608 (2007-03-27)
Felbamate is an antiepileptic drug that is associated with minimal toxicity in preclinical species such as rat and dog but has an unacceptable incidence of serious idiosyncratic reactions in man. Idiosyncratic reactions account for over half of toxicity-related drug failures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service