Skip to Content
Merck
All Photos(1)

Key Documents

F0652

Sigma-Aldrich

Anti-Factor IX antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Factor IX Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

dot blot: 1:8,000
indirect ELISA: 1:5,000
western blot: 1:3,000 using using reduced and non-reduced human plasma blots

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... F9(2158)

General description

Factor IX is a 55 kDa, single chain, vitamin K-dependent plasma zymogen. Factor IX is synthesized in liver parenchymal cells and requires a post-translational, vitamin K-dependent, modification in order to become a mature plasma zymogen. Factor IX concentration in human plasma ranges between 2.5-5 mg/ml and its half-life is ~24 hr. Human factor IX gene is about 40 kb in size and is localized at the distal end of the X-chromosome.
Four and a half LIM domains protein 1 is a protein encoded by the FHL1 gene in humans and is located on human chromosome Xq27.2. Factor IX (or Christmas factor) is one of the serine proteases of the coagulation system. It belongs to peptidase family S1 and its deficiency causes hemophilia B. The proteins belong to a novel family of LIM proteins that are expressed in human skeletal muscle. FHL1 gene is related to carcinogenesis and its inactivation is a frequent event during oral carcinogenesis. FHL1 downregulation in oral squamous cell carcinoma (OSCC) occurs through DNA methylation of the promoter region rather than histone deacetylation or mutation.

Specificity

Specifically reacts with human factor IX. The antibody detects human factor IX in both reduced and non-reduced normal human plasma.

Immunogen

human factor IX

Application

Anti-Factor IX antibody produced in rabbit has been used in immunoblotting and Gla enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Hereditary deficiencies or dysfunctions of factor IX cause hemophilia B or "Christmas Disease"(the surname of the first family described).A disulfide bond in factor IX connects the N-terminal sequence (light chain) of factor IX to the C-terminal sequence (heavy chain).Factor IX possesses higher sensitivity, hence even tiny amounts of residual intact can be visualized by F0652.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oguz Top et al.
Communications biology, 4(1), 964-964 (2021-08-14)
Production of biopharmaceuticals relies on the expression of mammalian cDNAs in host organisms. Here we show that the expression of a human cDNA in the moss Physcomitrium patens generates the expected full-length and four additional transcripts due to unexpected splicing.
Coagulation factor IX for hemophilia B therapy
Orlova NA, et al.
Acta Naturae (2012)
Hepatocyte Is a Sole Cell Type Responsible for the Production of Coagulation Factor IX In Vivo
Tatsumi K, et al.
Cell medicine, 3(1-3), 25-25 (2012)
An intragenic deletion of the factor IX gene in a family with hemophilia B.
Chen SH, et al.
The Journal of Clinical Investigation, 76(6), 2161-2161 (1985)
Variants in FIX propeptide associated with vitamin K antagonist hypersensitivity: functional analysis and additional data confirming the common founder mutations
Pezeshkpoor B, et al.
Annals of Hematology, 97(6), 1061-1069 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service