Skip to Content
Merck
All Photos(3)

Documents

F0300

Sigma-Aldrich

Anti-Fibroblast Growth Factor Receptor-2, Cytoplasmic antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FGFR-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115-125 kDa (doublet)

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2-4 μg/mL using trypsin-digested, human tissue sections
western blot: 0.25-0.5 μg/mL using extracts/lysates of FGFR-2 transfected cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGFR2(2263)

General description

Fibroblast growth factors receptors (FGFRs) are members of the tyrosine kinase family of growth factor receptors. They are glycosylated 110-150 k Da proteins that are constructed of an extracellular ligand binding region with either two or typically three immunoglobulin (Ig)-like domains and an eight amino acid ′acidic box′, a transmembrane region and a cytoplasmic split tyrosine kinase domain that is activated following ligand binding and receptor dimerization. At the mRNA level, FGFR-2 is highly expressed in developing human tissues including the brain (preferentially in glial cells), choroid plexus, skin, lung, kidney and bone. It is widely expressed in many adult human and animal tissues. It may be found in several anchorage-dependent cells, such as normal and malignant breast cancer cells.

Immunogen

synthetic peptide corresponding to amino acids 809-821 of the cytoplasmic region of human FGFR-2 (bek).

Application

Anti-Fibroblast Growth Factor Receptor-2 has been used in immunoblotting, immunoprecipitation, immunohistochemistry,western blotting.
Anti-Fibroblast Growth Factor Receptor-2, Cytoplasmic antibody produced in rabbit was used for immunohistochemistry of free-floating rat brain tissue sections.

Biochem/physiol Actions

Fibroblast growth factors (FGFs) and their receptors, FGFRs, are important regulators of cell proliferation, migration, tissue repair and differentiation during vertebrate development. FGFR-2 isoforms are expressed exclusively on cells of epithelial origin and are involved in tumor progression in thyroid cancers. Abnormalities in FGFR-2 expressions have been observed in skin, bone and cancer pathologies.
Fibroblast growth factors receptors (FGFRs) mediated signal transduction requires dimerization or oligomerization and autophosphorylation of the receptors through their tyrosine kinase domain. Mutations in the extracellular domain of FGFR-2 causes Crouzon, as well as other craniosynostosis syndromes (e.g. Pfeiffer′s, Apert′s, Jackson-Weiss′), disorders of human skeletal development.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative Caveats of Standard Immunohistochemical Procedures: Implications for Optical Disector-based Designs
Melvin N R. and Sutherland R J.
The Journal of Histochemistry and Cytochemistry, 58(7), 577-584 (2009)
Adriano Redler et al.
PloS one, 8(8), e72224-e72224 (2013-08-27)
Fibroblast growth factor receptor-2 (FGFR-2) plays an important role in tumorigenesis. In thyroid cancer it has been observed a FGFR-2 down-modulation, but the role of this receptor has not been yet clarified. Therefore, we decided to examine the expression of
Fibroblast growth factor receptor 2 (FGFR2)-mediated reciprocal regulation loop between FGF8 and FGF10 is essential for limb induction
Xu X, et al.
Development, 125(4), 753-765 (1998)
Masaru Katoh
The Journal of investigative dermatology, 129(8), 1861-1867 (2009-04-24)
Fibroblast growth factor receptor (FGFR)2 is regulated on the basis of the balance of FGFs, heparan-sulfate proteoglycans, FGFR2 isoforms, endogenous inhibitors, and microRNAs. FGFR2 signals cross-talk with hedgehog, bone morphogenetic protein, and other regulatory networks. Some cases of congenital skeletal
ERK signal suppression and sensitivity to CH5183284/Debio 1347, a selective FGFR inhibitor
Nakanishi Y, et al.
Molecular Cancer Therapeutics, 14(12), 2831-2839 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service