Skip to Content
Merck
All Photos(1)

Key Documents

EP5820747005

Caps for Eppendorf® S-4-104, S-4x1000 Round Buckets and S-4x750 Rotors

aerosol-tight, aerosol-tight, for 5810 / 5810R / 5910R / 5920R

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913
NACRES:
NB.82
Pricing and availability is not currently available.

feature

aerosol-tight

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5820747005 (Global)

technique(s)

centrifugation: suitable

compatibility

for use with 5810 R
for use with 5810
for use with 5910 R
for use with 5920 R

Features and Benefits

  • Ensures safe centrifugation of hazardous samples
  • Aerosol-tight caps ensure reduced air resistance of the rotor
  • Autoclavable

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service