Skip to Content
Merck
All Photos(1)

Key Documents

EP0030601700

Sigma-Aldrich

Eppendorf® Microplate 96/F-PP

PCR clean, flat bottom, black wells, white border, pkg of 80 ea (5 bags of 16 plates)

Synonym(s):

Microplate 96/F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.02
material:
black (with white border)
black wells
cap (push fit)
flat bottom
polycarbonate frame
polypropylene
polypropylene wells
size:
96 well
sterility:
non-sterile
feature:
PCR clean
RNase and DNase free

material

black (with white border)
black wells
cap (push fit)
flat bottom
polycarbonate frame
polypropylene
polypropylene wells

sterility

non-sterile

feature

PCR clean
RNase and DNase free

packaging

pkg of 80 ea (5 bags of 16 plates)

manufacturer/tradename

Eppendorf® 0030601700

technique(s)

PCR: suitable

capacity

1.5 mL

size

96 well

color

black wells
white border

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service