Skip to Content
Merck
All Photos(1)

Key Documents

EP0030504305

Eppendorf® Deepwell plates, Protein LoBind, 96 wells

white plate, conical bottom colorless wells, capacity 2000 μL, pkg of 20 ea

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom colorless wells
white plate
size:
96 wells
sterility:
non-sterile
binding type:
low binding surface
Pricing and availability is not currently available.

material

conical bottom colorless wells
white plate

sterility

non-sterile

packaging

pkg of 20 ea

manufacturer/tradename

Eppendorf® 0030504305

technique(s)

PCR: suitable

capacity

2000 μL

size

96 wells

working volume

2 mL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® Deepwell plates, Protein LoBind, 96 wells, white plate of 2000 μL capacity are available as PCR clean purity grade, free of human DNA, DNase, RNase and PCR inhibitors.
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • These plates maximize sample recovery by significantly reducing sample binding to the surface.
  • High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface guarantee reliable sealing in plates.

Maximize your protein sample recovery with Eppendorf Protein LoBind Plates. Eppendorf LoBind Plates are equipped with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for protein research or sensitive testing methods in proteomics, where they enable excellent results due to less sample binding to the plate surface.

Application

  • Preparation or storage of protein, peptide or antibody samples
  • Enzymatic assays – the hydrophilic surface reduces denaturation when interacting with the vessel wall
  • Avoiding sample loss when storing virus stock solutions
  • Storage of cell suspensions

Features and Benefits

  • Eppendorf LoBind material ensures outstanding sample recovery for better test results
  • Without surface coating (e.g. silicone) to minimize the risk of sample interference
  • Batch-certified purity grade PCR clean: free from human DNA, DNase, RNase and PCR inhibitors
  • Available as reaction vessel, microplate and deepwell plate for easy scaling
  • Unique high-contrast OptiTrack matrix: up to 30% faster sample detection and fewer pipetting errors
  • RecoverMax well design: Optimized well geometry for minimized residual or dead volume and excellent mixing properties
  • Raised corrugated edges and a smooth surface ensure reliable sealing of the plates

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service