Skip to Content
Merck
All Photos(1)

Key Documents

EHU221311

Sigma-Aldrich

MISSION® esiRNA

targeting human POU5F1 (1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAAAGCAGAAACCCTCGTGCAGGCCCGAAAGAGAAAGCGAACCAGTATCGAGAACCGAGTGAGAGGCAACCTGGAGAATTTGTTCCTGCAGTGCCCGAAACCCACACTGCAGCAGATCAGCCACATCGCCCAGCAGCTTGGGCTCGAGAAGGATGTGGTCCGAGTGTGGTTCTGTAACCGGCGCCAGAAGGGCAAGCGATCAAGCAGCGACTATGCACAACGAGAGGATTTTGAGGCTGCTGGGTCTCCTTTCTCAGGGGGACCAGTGTCCTTTCCTCTGGCCCCAGGGCCCCATTTTGGTACCCCAGGCTATGGGAGCCCTCACTTCACTGCACTGTACTCCTCGGTCCCTTTCCCTGAGGGGGAAGCCTTTCCCCCTGTCTCTGTCACCACTCTGGGCTCTCCCATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Liu et al.
Biochemical and biophysical research communications, 461(3), 525-532 (2015-04-26)
Our previous study showed that Octamer-binding transcription factor 4 (OCT4) expression was upregulated and significantly associated with histological grade through the analysis of OCT4 expression in 159 ovarian cancer tissue samples, and OCT4 mediated follicle-stimulating hormone (FSH)-induced anti-apoptosis in epithelial
Prathap Kumar S Mahalingaiah et al.
Journal of cellular physiology, 230(8), 1916-1928 (2014-12-30)
Oxidative injury to cellular macromolecules has been suggested as a common pathway shared by multiple etiological factors for kidney cancer. Whether the chronic oxidative stress alone is sufficient to induce malignant transformation in human kidney cells is not clear. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service