Skip to Content
Merck
All Photos(1)

Key Documents

EHU130711

Sigma-Aldrich

MISSION® esiRNA

targeting human FASN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTGGTCTTGAACTCCTTGGCGGAAGAGAAGCTGCAGGCCAGCGTGAGGTGCTTGGCTACGCACGGTCGCTTCCTGGAAATTGGCAAATTCGACCTTTCTCAGAACCACCCGCTCGGCATGGCTATCTTCCTGAAGAACGTGACATTCCACGGGGTCCTACTGGATGCGTTCTTCAACGAGAGCAGTGCTGACTGGCGGGAGGTGTGGGCGCTTGTGCAGGCCGGCATCCGGGATGGGGTGGTACGGCCCCTCAAGTGCACGGTGTTCCATGGGGCCCAGGTGGAGGACGCCTTCCGCTACATGGCCCAAGGGAAGCACATTGGCAAAGTCGTCGTGCAGGTGCTTGCGGAGGAGCCGGAGGCAGTGCTGAAGGGGGCCAAACCCAAGCTGATGTCGGCCATCTCCAAGACCTTCTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaokun Gang et al.
The Prostate, 79(8), 864-871 (2019-04-08)
Fatty acid synthase (FASN) is vital for maintaining lipid homeostasis in prostate cancer (PCa) cells, which have an increased rate of de novo fatty acid (FA) synthesis. Mutations in the gene encoding the tumor suppressor speckle-type POZ protein (SPOP), which
Xing Zhong et al.
Oncology letters, 21(4), 245-245 (2021-03-06)
Diffuse large B-cell lymphoma (DLBCL) is the most common and heterogeneous lymphoid malignancy. The subtype with MYC and BCL-2 double-expressor lymphoma (DEL) was defined by its aggressive nature and poor survival outcome. Therefore, the development of effective therapies for the
Arif Khan et al.
International journal of nanomedicine, 15, 5575-5589 (2020-08-18)
The overexpression of Her-2 in 25-30% breast cancer cases and the crosstalk between Her-2 and fatty acid synthase (FASN) establishes Her-2 as a promising target for site-directed delivery. The present study aimed to develop the novel lipid base formulations to
Valeryia Mikalayeva et al.
International journal of molecular sciences, 20(6) (2019-03-21)
Both cytosolic fatty acid synthesis (FAS) and mitochondrial fatty acid oxidation (FAO) have been shown to play a role in the survival and proliferation of cancer cells. This study aimed to confirm experimentally whether FAS and FAO coexist in breast
Débora C Bastos et al.
The Journal of pathology, 253(3), 292-303 (2020-11-10)
Loss of the tumor suppressor gene Pten in murine prostate recapitulates human carcinogenesis and causes stromal proliferation surrounding murine prostate intraepithelial neoplasia (mPIN), which is reactive to microinvasion. In turn, invasion has been shown to be regulated in part by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service