Skip to Content
Merck
All Photos(1)

Key Documents

EHU110531

Sigma-Aldrich

MISSION® esiRNA

targeting human PPIB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGTAGGCCGGGTGATCTTTGGTCTCTTCGGAAAGACTGTTCCAAAAACAGTGGATAATTTTGTGGCCTTAGCTACAGGAGAGAAAGGATTTGGCTACAAAAACAGCAAATTCCATCGTGTAATCAAGGACTTCATGATCCAGGGCGGAGACTTCACCAGGGGAGATGGCACAGGAGGAAAGAGCATCTACGGTGAGCGCTTCCCCGATGAGAACTTCAAACTGAAGCACTACGGGCCTGGCTGGGTGAGCATGGCCAACGCAGGCAAAGACACCAACGGCTCCCAGTTCTTCATCACGACAGTCAAGACAGCCTGGCTAGATGGCAAGCATGTGGTGTTTGGCAAAGTTCTAGAGGGCATGGAGGTGGTGCGGAAGGTGGAGAGCACCAAGACAGACAGCCGGGATAAACCCCTGAAGGATGTGATCATCGCAGACTGCGGCAAGATCGAGGTGGAGAAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Hanning et al.
British journal of cancer, 108(2), 450-460 (2013-01-10)
When designing therapeutic short-interfering RNAs (siRNAs), off-target effects (OTEs) are usually predicted by computational quantification of messenger RNAs (mRNAs) that contain matches to the siRNA seed sequence in their 3' UTRs. It is assumed that the higher the number of
Maria M Caffarel et al.
The Journal of pathology, 231(2), 168-179 (2013-06-15)
Oncostatin M receptor (OSMR) is commonly over-expressed in advanced cervical squamous cell carcinoma (SCC), producing a significantly worse clinical outcome. Cervical SCC cells that over-express OSMR show enhanced responsiveness to the major ligand OSM, which induces multiple pro-malignant effects, including
Paulo C M Urbano et al.
Frontiers in immunology, 10, 3047-3047 (2020-02-11)
Maintenance of regulatory T cells CD4+CD25highFOXP3+ (Treg) stability is vital for proper Treg function and controlling the immune equilibrium. Treg cells are heterogeneous and can reveal plasticity, exemplified by their potential to express IL-17A. TNFα-TNFR2 signaling controls IL-17A expression in
Ma Paz Zafra et al.
PloS one, 9(3), e91996-e91996 (2014-03-19)
Suppresors of cytokine signaling (SOCS) proteins regulate cytokine responses and control immune balance. Several studies have confirmed that SOCS3 is increased in asthmatic patients, and SOCS3 expression is correlated with disease severity. The objective of this study was to evaluate
T W Laetsch et al.
Cell death & disease, 5, e1072-e1072 (2014-02-22)
Cancer treatments induce cell stress to trigger apoptosis in tumor cells. Many cancers repress these apoptotic signals through alterations in the Bcl2 proteins that regulate this process. Therapeutics that target these specific survival biases are in development, and drugs that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service