Skip to Content
Merck
All Photos(1)

Key Documents

EHU091841

Sigma-Aldrich

MISSION® esiRNA

targeting human INSR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGTGCATCCCTGAGTGTCCCTCCGGGTACACGATGAATTCCAGCAACTTGCTGTGCACCCCATGCCTGGGTCCCTGTCCCAAGGTGTGCCACCTCCTAGAAGGCGAGAAGACCATCGACTCGGTGACGTCTGCCCAGGAGCTCCGAGGATGCACCGTCATCAACGGGAGTCTGATCATCAACATTCGAGGAGGCAACAATCTGGCAGCTGAGCTAGAAGCCAACCTCGGCCTCATTGAAGAAATTTCAGGGTATCTAAAAATCCGCCGATCCTACGCTCTGGTGTCACTTTCCTTCTTCCGGAAGTTACGTCTGATTCGAGGAGAGACCTTGGAAATTGGGAACTACTCCTTCTATGCCTTGGACAACCAGAACCTAAGGCAGCTCTGGGACTGGAGCAAACACAACCTCACCATCACTCAGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masafumi Aoki et al.
PloS one, 14(10), e0223528-e0223528 (2019-10-04)
The aim of this study was to identify changes in skin function associated with obesity and the mechanisms underlying these changes. Functional changes and gene expression in skin were investigated in C57BL/6J mice fed either a control or high-fat diet
Shingo Ito et al.
Neurochemistry international, 101, 22-29 (2016-11-05)
We have previously shown in SH-SY5Y human neuroblastoma cells that the expressions of basal (75 kDa) and high molecular weight (HMW; 85 kDa) isoforms of the p75 neurotrophic receptor (p75NTR) are stimulated by amyloid-β peptide1-42 oligomers (AβOs) via the insulin-like growth factor-1
Phoebe L Sarkar et al.
Frontiers in endocrinology, 10, 481-481 (2019-08-06)
Androgen deprivation therapy (ADT) is the standard treatment for advanced prostate cancer (PCa), yet many patients relapse with lethal metastatic disease. With this loss of androgens, increased cell plasticity has been observed as an adaptive response to ADT. This includes
Prasenjit Manna et al.
Archives of biochemistry and biophysics, 615, 22-34 (2017-01-09)
This study examined the hypothesis that vitamin-D prevents oxidative stress and upregulates glucose metabolism via activating insulin-independent signaling molecules in 3T3-L1 adipocytes and in high fat diet (HFD)-fed mice. To investigate the mechanism 3T3L1 adipocytes were treated with high glucose
Sandip Mukherjee et al.
PloS one, 12(1), e0169809-e0169809 (2017-01-11)
Dramatic increase of diabetes over the globe is in tandem with the increase in insulin requirement. This is because destruction and dysfunction of pancreatic β-cells are of common occurrence in both Type1 diabetes and Type2 diabetes, and insulin injection becomes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service