Skip to Content
Merck
All Photos(1)

Documents

EHU083661

Sigma-Aldrich

MISSION® esiRNA

targeting human WT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGGGCATGTGTATGTGTCTGCTAATGTAAACTTTGTCATGGTTTCCATTTACTAACAGCAACAGCAAGAAATAAATCAGAGAGCAAGGCATCGGGGGTGAATCTTGTCTAACATTCCCGAGGTCAGCCAGGCTGCTAACCTGGAAAGCAGGATGTAGTTCTGCCAGGCAACTTTTAAAGCTCATGCATTTCAAGCAGCTGAAGAAAAAATCAGAACTAACCAGTACCTCTGTATAGAAATCTAAAAGAATTTTACCATTCAGTTAATTCAATGTGAACACTGGCACACTGCTCTTAAGAAACTATGAAGATCTGAGATTTTTTTGTGTATGTTTTTGACTCTTTTGAGTGGTAATCATATGTGTCTTTATAGATGTACATACCTCCTTGCACAAATGGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Wang et al.
Theriogenology, 148, 8-17 (2020-03-04)
To determine the role of 3, 3', 5-triiodo-L thyroxine (T3) in the differentiation of Sertoli cells (SCs) and the factors influencing maturity via the Wilms' tumor 1 (WT1)/non-canonical Wnt signaling pathway, high purity SCs were isolated from newborn calves' testes
Tove Ullmark et al.
Biochemical and biophysical research communications, 482(4), 802-807 (2016-11-28)
Wilms' tumor gene 1 (WT1) is a zinc finger transcription factor that has been implicated as an oncogene in leukemia and several other malignancies. When investigating possible gene expression network partners of WT1 in a large acute myeloid leukemia (AML)
Junjie Chen et al.
Journal of experimental & clinical cancer research : CR, 35(1), 173-173 (2016-11-09)
The metastatic cascade is a complex and multistep process with many potential barriers. Recently, miR-193a has been reported to be a suppressive miRNA in multiple types of cancers, but its underlying anti-oncogenic activity in non-small cell lung cancers (NSCLC) is
Xue Wang et al.
Reproduction, fertility, and development, 32(5), 522-530 (2020-02-06)
The gap junction protein connexin (Cx) 43 between adjacent Sertoli cells (SCs) is the main testicular factor regulating the growth and development of SCs, and plays a vital role in controlling cell differentiation and maturation. However, the endogenous testicular factors
Paramahamsa Maturu et al.
Neoplasia (New York, N.Y.), 19(3), 237-249 (2017-03-04)
Wilms' tumors (WT), which accountfor 6% of all childhood cancers, arise from dysregulated differentiation of nephrogenic progenitor cells from embryonic kidneys. Though there is an improvement in the prognosis of WT, still 10% of patients with WT die due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service