Skip to Content
Merck
All Photos(1)

Key Documents

EHU043571

Sigma-Aldrich

MISSION® esiRNA

targeting human YY1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTCACCATGTGGTCCTCAGATGAAAAAAAAGATATTGACCATGAGACAGTGGTTGAAGAACAGATCATTGGAGAGAACTCACCTCCTGATTATTCAGAATATATGACAGGAAAGAAACTTCCTCCTGGAGGAATACCTGGCATTGACCTCTCAGATCCCAAACAACTGGCAGAATTTGCTAGAATGAAGCCAAGAAAAATTAAAGAAGATGATGCTCCAAGAACAATAGCTTGCCCTCATAAAGGCTGCACAAAGATGTTCAGGGATAACTCGGCCATGAGAAAACATCTGCACACCCACGGTCCCAGAGTCCACGTCTGTGCAGAATGTGGCAAAGCTTTTGTTGAGAGTTCAAAACTAAAACGACACCAACTGGTTCATACTGGAGAGAAGCCCTTTCAGTGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinpiao Lin et al.
Journal of autoimmunity, 77, 67-75 (2016-11-11)
Previous studies have revealed a critical role of YY1, a "Yin Yang" transcription factor, in cancer development and progression. However, whether YY1 has any role in rheumatoid arthritis (RA) remains unknown. This study aims to explore the potential role of
J-B Tian et al.
European review for medical and pharmacological sciences, 23(13), 5714-5729 (2019-07-13)
Increasing studies have confirmed long non-coding RNAs (lncRNAs) as novel regulators in tumorigenesis. LncRNA DDX11 antisense RNA 1 (DDX11-AS1) has been found to be abnormally expressed in several tumors. In this work, we aimed to evaluate its expressions and functions
Huan Liu et al.
Scientific reports, 10(1), 20493-20493 (2020-11-26)
Angiogenesis is a physiological process for the formation of new blood vessels from the pre-existing vessels and it has a vital role in the survival and growth of neoplasms. During tumor angiogenesis, the activation of the gene transcriptions in vascular
Jiyeon Choi et al.
Nature communications, 11(1), 2718-2718 (2020-06-03)
Genome-wide association studies (GWAS) have identified ~20 melanoma susceptibility loci, most of which are not functionally characterized. Here we report an approach integrating massively-parallel reporter assays (MPRA) with cell-type-specific epigenome and expression quantitative trait loci (eQTL) to identify susceptibility genes/variants
Feng Liang et al.
Journal of cellular biochemistry, 120(10), 18415-18424 (2019-06-14)
Lacking of diagnostic and prognostic biomarkers is a significant reason for the poor prognosis of patients with triple-negative breast cancer (TNBC). MicroRNAs (miRNAs) have been discovered to engage in the tumorigenesis and development of TNBC. miR-5590-3p has been found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service