Skip to Content
Merck
All Photos(1)

Key Documents

EHU022531

Sigma-Aldrich

MISSION® esiRNA

targeting human ULK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGAACCTCGCCAAGTCTCAGACGCTGCTGGGGAAGGAAATCAAAATCCTGAAGGAACTGAAACATGAAAACATCGTGGCCCTGTACGACTTCCAGGAAATGGCTAATTCTGTCTACCTGGTTATGGAGTACTGCAACGGTGGGGACCTGGCCGACTACCTGCACGCCATGCGCACGCTGAGCGAGGACACCATCAGGCTCTTCCTGCAGCAGATCGCGGGCGCCATGCGGCTTCTGCACAGCAAAGGCATCATCCACCGCGACCTGAAACCGCAGAACATCCTGCTGTCCAACCCCGCCGGCCGCCGCGCCAACCCCAACAGCATCCGCGTCAAGATCGCTGACTTCGGCTTCGCGCGGTACCTCCAGAGCAACATGATGGCGGCCACACTCTGCGGCTCCCCCATGTACATGGCCCCCGAGGTCATCATGTCCCAGCACTACGACGGGAAGGCGGACCTGTGGAGCATCGGCACCATCGTCTACCAGTGCCTGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Spengler et al.
Cells, 9(3) (2020-03-15)
AMP-activated protein kinase (AMPK) is activated by vascular endothelial growth factor (VEGF) in endothelial cells and it is significantly involved in VEGF-induced angiogenesis. This study investigates whether the VEGF/AMPK pathway regulates autophagy in endothelial cells and whether this is linked
Min Liu et al.
Toxicology letters, 283, 106-115 (2017-11-13)
Mitochondrial aldehyde dehydrogenase 2 (ALDH2), an important enzyme in the elimination of toxic aldehydes, is involved in cardioprotection against diabetes mellitus. This study was designed to examine the mechanism behind ALDH2-offered protection against high glucose exposure with a focus on
Shan Zhu et al.
Autophagy, 9(3), 317-327 (2012-12-18)
IFN1@ (interferon, type 1, cluster, also called IFNα) has been extensively studied as a treatment for patients with chronic myeloid leukemia (CML). The mechanism of anticancer activity of IFN1@ is complex and not well understood. Here, we demonstrate that autophagy
Qiang Xiao et al.
Theranostics, 10(22), 10245-10261 (2020-09-16)
Hepatocellular carcinoma (HCC) is the third most frequent cause of cancer-related deaths globally because of high metastasis and recurrence rates. Elucidating the molecular mechanisms of HCC recurrence and metastasis and developing effective targeted therapies are expected to improve patient survival.
Tiejian Nie et al.
Cell death & disease, 7(12), e2563-e2563 (2016-12-30)
Endoplasmic reticulum (ER) stress is involved in many cellular processes. Emerging evidence suggests that ER stress can trigger autophagy; however, the mechanisms by which ER stress regulates autophagy and its role in this condition are not fully understood. HIV Tat-interactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service