Skip to Content
Merck
All Photos(1)

Key Documents

E7906

Sigma-Aldrich

Esomeprazole magnesium hydrate

≥98% (HPLC)

Synonym(s):

(S)-Omeprazole magnesium hydrate, (T-4)-Bis[6-methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl-κO]-1H-benzimidazolato-κN3]-Magnesium hydrate, Nexium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H36MgN6O6S2 · xH2O
CAS Number:
Molecular Weight:
713.12 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -128 to -146°, c = 0.5 in methanol

color

white to off-white

solubility

DMSO: ≥5 mg/mL

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

O.COc1ccc2n([Mg]n3c(nc4cc(OC)ccc34)S(=O)Cc5ncc(C)c(OC)c5C)c(nc2c1)S(=O)Cc6ncc(C)c(OC)c6C

InChI

1S/2C17H18N3O3S.Mg.H2O/c2*1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;;/h2*5-8H,9H2,1-4H3;;1H2/q2*-1;+2;/t2*24-;;/m00../s1

InChI key

LGRFYKXKCZKDLS-DIYDTZFBSA-N

Related Categories

General description

Esomeprazole is the (S)-isomer of omeprazole. It blocks the H+/K+-ATPase enzyme and lowers the release of hydrochloric acid by gastric parietal cells. Esomeprazole is used for treating gastroesophageal reflux disease (GERD).

Application

Esomeprazole magnesium hydrate has been used to induce intracellular acidification in an ATP12A-independent manner.

Biochem/physiol Actions

Esomeprazole magnesium dihydrate is a leading proton pump inhibitor.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esomeprazole increases airway surface liquid pH in primary Cystic Fibrosis epithelial cells
Delpiano L, et al.
Frontiers in Pharmacology, 9 (2018)
Yukiko Nakagawa et al.
Biological & pharmaceutical bulletin, 43(4), 682-687 (2020-04-03)
We previously showed that adhesive aggregates were formed when levofloxacin hydrate tablets and lansoprazole orally disintegrating (OD) tablets were suspended in water in the clinical context. In this study, we have clarified the factors causing aggregate formation, focusing on the
Esomeprazole
Scott LJ, et al.
Drugs, 62(10), 1503-1538 (2002)
Mark Rochman et al.
The Journal of allergy and clinical immunology, 147(5), 1924-1935 (2020-12-09)
Proton pump inhibitors (PPIs) have been recognized as a primary treatment of eosinophilic esophagitis (EoE), an allergic inflammatory disease of the esophageal mucosa. The mechanisms underlying esophageal epithelial responses to PPIs remain poorly understood. We hypothesized that PPIs can counteract
A review of esomeprazole in the treatment of gastroesophageal reflux disease (GERD)
Kalaitzakis E and Bjornsson E
Therapeutics and Clinical Risk Management, 3(4), 653-653 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service