Skip to Content
Merck
All Photos(1)

Documents

E7407

Sigma-Aldrich

ERK1, active, untagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAPK3, P44ERK1, P44MAPK, PRKM3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

329-445 nmol/min·mg

mol wt

~44 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPK3(5595)

General description

Extracellular signal-regulated kinase 1 (ERK1) belongs to mitogen-activated protein kinases (MAPK)/ERK family of intermediate proteins in the signaling pathways. This gene contains nine exons and eight introns, and spans 9kb. It is a constitutively expressed protein, the activity of which is altered by the phosphorylation state. It has a higher expression level in nervous tissues. It contains 11 kinase subdomains, common between humans and mice. It has a molecular weight of 44kDa.

Biochem/physiol Actions

ERK1 (MAPK3) participates in cellular signaling cascades that are activated in response to numerous growth factors and cytokines. Disruption of MAP kinase signaling functionality is associated with disease states including inflammation, cancer and neurodegenerative disease.
Extracellular signal-regulated kinase 1 (ERK1) act as intermediate in multiple signaling cascades, and is acted upon by MEK protein. MEK are essential for Golgi fragmentation during mitosis, and this is achieved by its action upon an alternatively spliced isoform of ERK1, called ERK1c. ERK1/2 signaling plays a key role in cellular response to topoisomerase II poisoning, by activating G2/M checkpoint, and thus, prevents poison-induced cell death. In melanocytes, pleiotrophin suppresses pigmentation or melanogenesis, by degradation of MITF, through the activation of ERK1/2 signaling.

Physical form

Supplied in 25 mM MOPS, pH 7.5, with 300 mM NaCl, 0.25 mM DTT, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel M Aebersold et al.
Molecular and cellular biology, 24(22), 10000-10015 (2004-10-29)
Extracellular signal-regulated kinases (ERKs) are signaling molecules that regulate many cellular processes. We have previously identified an alternatively spliced 46-kDa form of ERK1 that is expressed in rats and mice and named ERK1b. Here we report that the same splicing
F García et al.
Genomics, 50(1), 69-78 (1998-06-20)
The complete genomic structure of the human p44(mapk) gene (HMGW-approved symbol PRKM3) has been determined. The gene covers 9 kb and is composed of nine exons and eight introns. This structure is identical to the previously reported mouse p44(mapk) gene
Runping Fang et al.
Nature communications, 12(1), 177-177 (2021-01-10)
Glioblastoma (GBM) is the most common type of adult malignant brain tumor, but its molecular mechanisms are not well understood. In addition, the knowledge of the disease-associated expression and function of YTHDF2 remains very limited. Here, we show that YTHDF2
Ryan H Kolb et al.
PloS one, 7(11), e50281-e50281 (2012-11-21)
Topo II poisons, which target topoisomerase II (topo II) to generate enzyme mediated DNA damage, have been commonly used for anti-cancer treatment. While clinical evidence demonstrate a capability of topo II poisons in inducing apoptosis in cancer cells, accumulating evidence
Yoav D Shaul et al.
The Journal of cell biology, 172(6), 885-897 (2006-03-15)
Extracellular signal-regulated kinase 1c (ERK1c) is an alternatively spliced form of ERK1 that is regulated differently than other ERK isoforms. We studied the Golgi functions of ERK1c and found that it plays a role in MEK-induced mitotic Golgi fragmentation. Thus

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service