Skip to Content
Merck
All Photos(3)

Documents

E5391

Sigma-Aldrich

Ethylenediaminetetraacetic acid tetrasodium salt hydrate

≥99.0%

Synonym(s):

EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COONa)2 · xH2O
CAS Number:
Molecular Weight:
380.17 (anhydrous basis)
Beilstein:
3861753
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

biological source

synthetic (organic)

Quality Level

Assay

≥99.0%

form

powder

technique(s)

ligand binding assay: suitable

impurities

Insoluble matter, passes filter test

pH

11.0-12.0 (0.1 M in H2O)

solubility

H2O: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.01%

cation traces

Al: ≤0.001%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: <0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.06 at 280 in H2O at 0.1 M
≤1.5 at 260 in H2O at 0.1 M

application(s)

sample preparation

storage temp.

room temp

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.4Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;1H2/q;4*+1;/p-4

InChI key

GXFFSKMBMYHTAE-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

EDTA is an inhibitor of metalloproteases, at effective concentrations of 1-10 mM. EDTA acts as a chelator of the zinc ion in the active site of metalloproteases, and can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases. For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used. The optimal concentration is 1.5 mg per ml of blood.

Preparation Note

Soluble in water at room temperature up to 1.45 M. The pH of this solution will be approximately 10 to 11 and will be rather viscous.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J Smith et al.
STAR protocols, 4(1), 102002-102002 (2023-01-08)
Here, we provide a protocol using chemical pulldown combined with mass spectrometry (LC-MS/MS) to identify drug targets in Plasmodium falciparum. This approach works upon the principle that a resin-bound inhibitor selectively binds its molecular target(s) in cell-free lysates. We describe
Carl Marfurt et al.
Investigative ophthalmology & visual science, 60(15), 4972-4984 (2019-12-04)
The subbasal nerve plexus (SNP) is the densest and most recognizable component of the mammalian corneal innervation; however, the anatomical configuration of the SNP in most animal models remains incompletely described. The purpose of the current study is to describe
Philip Thomas et al.
Nature protocols, 4(6), 825-837 (2009-05-16)
The Buccal Micronucleus Cytome (BMCyt) assay is a minimally invasive method for studying DNA damage, chromosomal instability, cell death and the regenerative potential of human buccal mucosal tissue. This method is increasingly used in molecular epidemiological studies for investigating the
Analyzing the developmental expression of sigma factors with S1-nuclease mapping.
J Kormanec
Methods in molecular biology (Clifton, N.J.), 160, 481-494 (2001-03-27)
Arjeta Podrimaj-Bytyqi et al.
Scientific reports, 8(1), 17873-17873 (2018-12-16)
Bladder urothelial cell carcinoma (UCC) is an increasingly prevalent cancer worldwide, and thus, gaining a better understanding of its identifiable risk factors is a global priority. This study addressed this public health need with the understanding that cancer-initiating events, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service