Skip to Content
Merck

Skip To

E3652

Epoxomicin

≥95% (HPLC), solid

Synonym(s):

N-Acetyl-N-methyl-L-isoleucyl-L-isoleucyl-N-[(1S)-3-methyl-1-[[(2R)-2-methyloxiranyl]carbonyl]butyl]-L-threoninamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H]([C@@H](C)CC)N(C)C(C)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(C)C)C(=O)[C@@]1(C)CO1

InChI

1S/C28H50N4O7/c1-11-16(5)21(30-27(38)23(17(6)12-2)32(10)19(8)34)25(36)31-22(18(7)33)26(37)29-20(13-15(3)4)24(35)28(9)14-39-28/h15-18,20-23,33H,11-14H2,1-10H3,(H,29,37)(H,30,38)(H,31,36)/t16-,17-,18+,20-,21-,22-,23-,28+/m0/s1

InChI key

DOGIDQKFVLKMLQ-JTHVHQAWSA-N

General description

Epoxomicin is a linear peptide consisting of a threonine or serine residue with α′, β′-epoxyketone derived from leucine or a γ,δ-dehydroleucine. It is a natural product isolated from Actinomyces sp., and is a cell-permeable, potent, selective and irreversible proteasome inhibitor.

Application

Epoxomicin has been used:
  • as an ubiquitin–proteosome system (UPS) inhibitor in pheochromocytoma PC12 cells
  • as a proteasome inhibitor in mammary epithelial MCF-10A cells
  • as a proteasome inhibitor in chymotryptic assay in cardiomyocytes

Biochem/physiol Actions

Epoxomicin, a natural product isolated from Actinomyces sp., is a cell-permeable, potent, selective and irreversible proteasome inhibitor.
Epoxomicin binds covalently to the catalytic subunits of proteasome. It forms an adduct with target proteins. It inhibits chymotrypsin-like activity of the proteasome. Epoxomicin also inhibits the nuclear factor κ light chain enhancer of activated B cells (NF-κB) mediated proinflammatory signalling pathway. It is also a potent antitumor and anti-inflammatory agent.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EI8E9661E5161
assay

≥95% (HPLC)

assay

-

assay

≥95% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C

form

solid

form

-

form

-

form

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters