Skip to Content
Merck

E2128

β-Estradiol 3-(β-D-glucuronide) 17-sulfate dipotassium salt

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 3-glucuronide 17-sulfate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-glucuronide 17-sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C24H30O11SK2
CAS Number:
Molecular Weight:
604.75
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
MDL number:
Assay:
≥98% (TLC)
Form:
powder
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Level

assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

[K].CC12CCC3C(CCc4cc(OC5OC(C(O)C(O)C5O)C(O)=O)ccc34)C1CCC2OS(O)(=O)=O

InChI

1S/C24H32O11S.K.H/c1-24-9-8-14-13-5-3-12(33-23-20(27)18(25)19(26)21(34-23)22(28)29)10-11(13)2-4-15(14)16(24)6-7-17(24)35-36(30,31)32;;/h3,5,10,14-21,23,25-27H,2,4,6-9H2,1H3,(H,28,29)(H,30,31,32);;

InChI key

HHYDRCVKADTNKW-UHFFFAOYSA-N

Biochem/physiol Actions

Estradiol is derived from estrogen and it exists as a conjugate as sulfate and glucuronides. This conjugation enables in bringing down the estradiol hormonal activity and aids in eliminating them through excretion.


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B T Zhu et al.
Carcinogenesis, 19(1), 1-27 (1998-02-24)
Cytochrome P450 enzymes that metabolize estrogens are expressed in the mammary gland, uterus, brain and other target tissues for estrogen action, and this results in the formation of hydroxylated estrogens in these tissues. Estradiol metabolites formed in target tissues at
H Kuhl
Climacteric : the journal of the International Menopause Society, 8 Suppl 1, 3-63 (2005-08-23)
This review comprises the pharmacokinetics and pharmacodynamics of natural and synthetic estrogens and progestogens used in contraception and therapy, with special consideration of hormone replacement therapy. The paper describes the mechanisms of action, the relation between structure and hormonal activity



Global Trade Item Number

SKUGTIN
E2128-25MG04061832849836
E2128-5MG04061832849843