Skip to Content
Merck

E1877

Estriol 16α-(β-D-glucuronide)

≥97%

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol 16-glucuronide, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 16-glucuronide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H32O9
CAS Number:
Molecular Weight:
464.51
UNSPSC Code:
12352202
NACRES:
NA.77
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
67674
Assay:
≥97%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97%

form

powder

technique(s)

radioimmunoassay: suitable

solubility

ethanol: water (1:1): 10 mg/mL, clear to very slightly hazy, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)[C@@H](C[C@@]3([H])[C@]1([H])CCc4cc(O)ccc24)O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(O)=O

InChI

1S/C24H32O9/c1-24-7-6-13-12-5-3-11(25)8-10(12)2-4-14(13)15(24)9-16(21(24)29)32-23-19(28)17(26)18(27)20(33-23)22(30)31/h3,5,8,13-21,23,25-29H,2,4,6-7,9H2,1H3,(H,30,31)/t13-,14-,15+,16-,17+,18+,19-,20+,21+,23-,24+/m1/s1

InChI key

FQYGGFDZJFIDPU-JRSYHJKYSA-N



Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library