Skip to Content
Merck

E101

S-(−)-Eticlopride hydrochloride

≥98% (HPLC), D2 dopamine receptor antagonist, powder

Synonym(s):

FLB 131, S-(−)-3-Chloro-5-ethyl-N-[(1-ethyl-2-pyrrolidinyl)methyl]-6-hydroxy-2-methoxybenzamide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H25ClN2O3 · HCl
CAS Number:
Molecular Weight:
377.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

S-(−)-Eticlopride hydrochloride, ≥98% (HPLC), powder

Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]27/D −7.2°, c = 0.3 in methanol(lit.)

storage condition

desiccated

color

off-white

solubility

H2O: soluble >10 mg/mL, ethanol: soluble 2.6 mg/mL, 0.1 M HCl: soluble 7 mg/mL

SMILES string

Cl.CCN1CCC[C@H]1CNC(=O)c2c(O)c(CC)cc(Cl)c2OC

InChI

1S/C17H25ClN2O3.ClH/c1-4-11-9-13(18)16(23-3)14(15(11)21)17(22)19-10-12-7-6-8-20(12)5-2;/h9,12,21H,4-8,10H2,1-3H3,(H,19,22);1H/t12-;/m0./s1

InChI key

HFJFXXDHVWLIKX-YDALLXLXSA-N

Gene Information

human ... DRD2(1813)

Application

S-(−)-Eticlopride hydrochloride has been used in astrocyte-neuron coculture and functional magnetic resonance imaging (fMRI) experiments.

Biochem/physiol Actions

Potent and selective D2 dopamine receptor antagonist. Eticlopride is highly specific for DAD2 receptors than pimozide or haloperidol. The gene encoding the D2 dopamine receptor (DRD2) is linked with a several disorders including Parkinson′s disease, schizophrenia, and susceptibility to alcoholism.

Preparation Note

Solutions may be stored at 4° C for several days

Legal Information

Manufactured and sold with the permission of Astra AB.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library