Skip to Content
Merck
All Photos(2)

Key Documents

E0887

Sigma-Aldrich

Esterase from rabbit liver

lyophilized powder, ≥30 units/mg protein

Synonym(s):

Carboxyl esterase, Carboxylic-ester hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

specific activity

≥30 units/mg protein

mol wt

60 kDa

composition

Protein, ≥85% biuret

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Esterase belongs to the C-family of carboxylesterases. It is a glycoprotein, mainly present in the endoplasmic reticulum of hepatocytes.

Application

Esterase from rabbit liver has been used in a study to investigate a toxic effect of carbamate insecticides. Esterase from rabbit liver has also been used in a study to investigate the effect of simvastatin on expression and activity of a lipoprotein-associated phospholipase A(2).
Esterase from rabbit liver has been used:
  • in comparative biophysical studies with bovine bile-salt-activated lipase in the assays with vinyl acetate, triacylglycerols, p-nitrophenyl esters and serine enzyme inhibitors
  • in the catalysis and kinetic profiling studies of peptidic compounds
  • in in vitro stability assays of prodrug compounds (BMS-292655 and BMS300043)
  • in the hydrolysis of oleuropein aglycone (Ole-AG)

The enzyme from Sigma has been used to study the effect of divalent metal ions on the activity of esterase.

Biochem/physiol Actions

Esterase enzyme catalyzes hydrolysis of carboxylic ester to carboxylate and alcohol in the presence of water. Rabbit liver carboxylesterase catalyzes enantioselective hydrolysis of cineol derivatives.
Esterase enzyme has broad substrate specificity. Esterase from rabbit liver has been used to investigate the toxic effect of carbamate insecticides. Esterase from rabbit liver has also been used for the hydrolysis of simvastatin.

Unit Definition

One unit will hydrolyze 1.0 μmole of ethyl butyrate to butyric acid and ethanol per min at pH 8.0 at 25 °C.

Physical form

Lyophilized powder containing Tris buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shijin Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 36(22), 6261-6267 (2020-05-19)
To explore the potential of step-by-step assembly in the fabrication of biological materials, we designed and synthesized two peptide-based molecules for enzyme-instructed hierarchical assembly. Upon the treatment of alkaline phosphatase, one molecule undergoes enzyme-instructed self-assembly forming uniformed nanofibers. The other
Henri Chahinian et al.
Biochimica et biophysica acta, 1801(11), 1195-1204 (2010-07-27)
To differentiate esterases from lipases at the structure-function level, we have compared the kinetic properties and structural features of sequence-related esterase 1 from rabbit liver (rLE) and bile-salt-activated lipase from bovine pancreas (bBAL). In contrast to rLE, bBAL hydrolyses water-insoluble
Hansjörg Schwertz et al.
Journal of cardiovascular pharmacology, 52(2), 151-160 (2008-08-02)
Animal data strongly support a role for inflammation in myocardial ischemia reperfusion injury. Attempts at cardioprotection by immunomodulation (such as with the specific C5 antibody pexelizumab) in humans have been disappointing. We hypothesized that a broader spectrum antiinflammatory agent might
Z Qiao et al.
The Journal of international medical research, 37(4), 1029-1037 (2009-09-19)
Lipoprotein-associated phospholipase A(2) (Lp-PLA(2)) contributes to atherosclerotic plaque instability and subsequent sudden coronary death. Statins are associated with decreased stroke risk and may improve stability of atherosclerotic plaques. The present study investigated the effect of simvastatin on expression of Lp-PLA(2)
Anna Sobeková et al.
Acta biologica Hungarica, 60(1), 45-54 (2009-04-22)
The activities of the antioxidant and detoxifying enzymes, superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GSHPx), glutathione reductase (GR), glutathione-S-transferase (GST), and the content of thiobarbituric acid reactive substances (TBARS) were determined in the liver and kidney of rabbits after

Protocols

Objective: To standardize a procedure for the enzymatic determination of Esterase activity using Ethyl Butyrate as a substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service