Skip to Content
Merck
All Photos(1)

Key Documents

D8517

Sigma-Aldrich

Iron–Dextran

95-105 mg/mL (Iron content), solution

Synonym(s):

Ferric hydroxide dextran complex

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

synthetic

Quality Level

sterility

aseptically filled

Assay

95-105 mg/mL (Iron content)

form

solution

contains

0.5% phenol as preservative

composition

Iron, ~100 mg/mL

color

brown

useful pH range

4 - 6.5

storage temp.

room temp

SMILES string

[Fe].[S](=O)(=O)(O)O

InChI

1S/Fe.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)

InChI key

MVZXTUSAYBWAAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Iron-Dextran (ferric hydroxide dextran complex) may be used as an intravenous iron delivery preparation. Iron-Dextran may be used to induce iron-overload to study its effects and preventative measures.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Won Cho et al.
Sleep medicine, 14(3), 274-277 (2013-01-22)
Various techniques used to assess brain iron concentrations have demonstrated the presence of low iron stores in patients with restless legs syndrome (RLS). Previous open-label and randomized studies generally support the value of iron treatment for RLS symptoms. Only one
John C Wood et al.
Magnetic resonance in medicine, 60(1), 82-89 (2008-06-27)
MRI is gaining increasing importance for the noninvasive quantification of organ iron burden. Since transverse relaxation rates depend on iron distribution as well as iron concentration, physiologic and pharmacologic processes that alter iron distribution could change MRI calibration curves. This
Markus R Jahn et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 78(3), 480-491 (2011-03-29)
The treatment of iron deficiency anemia with polynuclear iron formulations is an established therapy in patients with chronic kidney disease but also in other disease areas like gastroenterology, cardiology, oncology, pre/post operatively and obstetrics' and gynecology. Parenteral iron formulations represent
D L Burns et al.
Nutrition (Burbank, Los Angeles County, Calif.), 11(2), 163-168 (1995-03-01)
Iron dextran was introduced more than 30 yr ago for the parenteral treatment of iron deficiency anemia that is refractory to oral therapy. Iron dextran is a preparation of ferric hydroxide complexed with a low molecular weight fraction of dextran.
Priya Prakash Karmali et al.
Molecular pharmaceutics, 9(3), 539-545 (2012-01-17)
Premature recognition and clearance of nanoparticulate imaging and therapeutic agents by macrophages in the tissues can dramatically reduce both the nanoparticle half-life and delivery to the diseased tissue. Grafting nanoparticles with hydrogels prevents nanoparticulate recognition by liver and spleen macrophages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service