Skip to Content
Merck
All Photos(3)

Key Documents

D8515

Sigma-Aldrich

Deoxyribonucleic acid−cellulose double-stranded from calf thymus DNA

lyophilized powder

Synonym(s):

DNA, Deoxyribonucleic acid, ds-DNA-Cellulose, DNA-Cellulose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
NACRES:
NA.51

form

lyophilized powder

extent of labeling

3-8 mg double-stranded calf thymus DNA/g

storage temp.

−20°C

General description

Deoxyribonucleic acid−cellulose double-stranded from calf thymus DNA, also known as ctDNA is a natural source of DNA designed for DNA binding agent studies. The detection limits of ctDNA are around 2.0ng and the linear ranges of light scattering intensity (I(LS)) is 0.05-1.5 mµg.

Application

Deoxyribonucleic acid−cellulose double-stranded from calf thymus DNA is used to study the binding parameters and binding specificity of a variety of proteins, nucleotides, and chemical reagents including anti-cancer agents. It is frequently used in pull-down assays and chromatographic procedures.

Quality

May contain a trace of Tris and EDTA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Zhou et al.
Journal of immunology (Baltimore, Md. : 1950), 198(7), 2568-2577 (2017-02-22)
CD74 mediates MHC class-II antigenic peptide loading and presentation and plays an important role in the pathogenesis of autoimmune diseases, including systemic lupus erythematosus. C57BL/6
Lutz Wobbe et al.
Nucleic acids research, 41(13), 6553-6567 (2013-05-08)
The molecular function of mTERFs (mitochondrial transcription termination factors) has so far only been described for metazoan members of the protein family and in animals they control mitochondrial replication, transcription and translation. Cells of photosynthetic eukaryotes harbour chloroplasts and mitochondria
A Georgaki et al.
Nucleic acids research, 22(7), 1128-1134 (1994-04-11)
A DNA helicase from calf thymus, called DNA helicase F, copurified with replication protein A through several steps of purification including DEAE-Sephacel, hydroxyapatite and single stranded DNA cellulose. It is finally separated from replication protein A on FPLC Mono Q
D Pisetsky et al.
The Journal of rheumatology, 26(9), 1934-1938 (1999-09-24)
To elucidate the epitope structure to DNA by identifying antigenic determinants on bacterial DNA bound by anti-DNA antibodies from normal human subjects (NHS) and patients with systemic lupus erythematosus (SLE). Sera from NHS and patients with SLE were tested by
T Kuzuhara et al.
Biochemical and biophysical research communications, 362(4), 805-810 (2007-09-04)
Tumor necrosis factor-alpha (TNF-alpha) inducing protein (Tipalpha) is a carcinogenic factor secreted from Helicobacter pylori (H. pylori), mediated through both enhanced expression of TNF-alpha and chemokine genes and activation of nuclear factor-kappaB. Since Tipalpha enters gastric cancer cells, the Tipalpha

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service