Skip to Content
Merck
All Photos(2)

Documents

D7384

Sigma-Aldrich

Decanal

≥98% (GC), liquid

Synonym(s):

Caprinaldehyde, Decyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CHO
CAS Number:
Molecular Weight:
156.27
Beilstein:
1362530
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.25

vapor density

>1 (vs air)

vapor pressure

~0.15 mmHg ( 20 °C)

Assay

≥98% (GC)

form

liquid

color

clear colorless

refractive index

n20/D 1.428 (lit.)

bp

207-209 °C (lit.)

density

0.83 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCCCCCCC

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h10H,2-9H2,1H3

InChI key

KSMVZQYAVGTKIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • The enhancement of flowery-like aroma in green tea under optimized processing conditions by sensory-directed flavor analysis.: This paper discusses how optimized processing conditions enhance the flowery-like aroma in green tea, with decanal being one of the key aromatic compounds, relevant to flavor chemistry and biochemistry (Wang et al., 2024).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Dekker et al.
Food additives and contaminants, 20(2), 180-185 (2003-03-08)
One of the phenomena in food-packaging interactions is flavour absorption. Absorption of flavour compounds from food products into food-packaging materials can result in loss of flavour compounds or an unbalance in the flavour profile changing a product's quality. The food
Anolda Cetkauskaite et al.
Environmental toxicology, 19(4), 372-386 (2004-07-23)
The aim of this research was to analyze the effects and the modes of action of elemental sulfur (S(0)) in bioluminescence and respiration of Vibrio fischeri cells and the enzymes crude luciferase, pure catalase, and alcohol dehydrogenase (ADH). Metallic copper
Giovanna Romano et al.
The Journal of experimental biology, 206(Pt 19), 3487-3494 (2003-08-27)
The diatom-derived aldehyde 2-trans-4-trans-decadienal (DD) was tested as an apoptogenic inducer in both copepod and sea urchin embryos, using terminal-deoxynucleotidyl-transferase-mediated dUTP nick-end labelling (TUNEL), DNA fragmentation profiling (laddering) and an assay for caspase-3 activity. DD induced TUNEL positivity and DNA
K Hör et al.
Journal of agricultural and food chemistry, 49(1), 21-25 (2001-02-15)
Based on (2)H/(1)H ratio measurements of commercial synthetic and "natural" references, the recently developed on-line gas chromatography pyrolysis isotope ratio mass spectrometry (HRGC-P-IRMS) technique was used to determine the delta(2)H(SMOW) values of the flavor compounds decanal, linalool, and linalyl acetate
R W G Van Willige et al.
Food additives and contaminants, 20(1), 84-91 (2003-01-10)
The influence of flavour absorption by low-density polyethylene (LDPE), polycarbonate (PC) and polyethylene terephthalate (PET) on taste perception of a model solution containing seven flavour compounds and orange juice in glass bottles was studied with and without pieces of the

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service