Skip to Content
Merck
All Photos(1)

Key Documents

D4941

Sigma-Aldrich

Anti-phospho-DAP-Kinase (pSer308) antibody, Mouse monoclonal

clone DKPS308, purified from hybridoma cell culture

Synonym(s):

Anti-DAPK, Anti-ROCO3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

DKPS308, monoclonal

form

buffered aqueous solution

mol wt

antigen ~160 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1-2 μg/mL using 293T (human embryonal kidney) cells transfected with DAP-kinase expression vector.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer308)

Gene Information

human ... DAPK1(1612)

General description

Monoclonal Anti-phospho DAP-Kinase (pSer308) (mouse IgG1 isotype) is derived from the DKPS308 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a phosphopeptide of human DAP-kinase, conjugated to KLH. Death Associated Protein Kinase (DAPK) is characterized with a multidomain structure including subdomain typical of serine/threonine kinases, a Ca2+/calmodulin regulatory domain, eight ankyrin repeats followed by two P-loop motifs and a typical death domain module. In addition, it also contains two auto-inhibitory domains, one of them Ca2+/calmodulin dependent. In the absence of this latter domain, DAPK is constitutively active.

Immunogen

Phosphopeptide corresponding to amino acids 303-312 (pSer308) of human DAP-kinase, conjugated to KLH.

Application

Anti-phospho-DAP-Kinase (pSer308) antibody, Mouse monoclonal has been used in:
  • western blot assay
  • immunocytochemistry
  • enzyme linked immunosorbent assay (ELISA)
  • immunoprecipitation

Monoclonal anti-phospho-DAP-kinase (pSer308) antibody can be used in indirect ELISA, immunoblotting and immunoprecipitation. It can also be used in western blotting.

Biochem/physiol Actions

Death Associated Protein Kinase (DAPK) plays crucial role in programmed cell death as well as in autophagy. DAPK activity is regulated by phosphorylation. Autophosphorylation at Ser308 on the calmodulin regulatory domain negatively regulate DAPK activity. This autophosphorylation, which occurs in cells at the basal state, lowers the affinity of DAPK for calmodulin and thus the kinase is inactive. Under some apoptotic conditions DAPK undergoes dephosphorylation. Consequently, it binds to calmodulin with higher affinity, becomes activated, phosphorylates its downstream substrate proteins, and mediates apoptosis. Monoclonal anti-phospho-DAP-kinase (pSer308) antibody can be used to study the mechanism of DAPK activation in apoptosis. It can also be used in microarray.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of DAPK-BimEL pathway in neuronal death induced by oxygen-glucose deprivation
He C, et al.
Neuroscience, 258, 254-254 (2014)
Xiao-Hui Li et al.
Neural regeneration research, 18(8), 1795-1801 (2023-02-09)
We previously showed that death-associated protein kinase 1 (DAPK1) expression is increased in hippocampal tissue in a mouse model of major depressive disorde and is related to cognitive dysfunction in Alzheimer's disease. In addition, depression is a risk factor for
The Pro-apoptotic Function of Death-associated Protein Kinase Is Controlled by a Unique Inhibitory Autophosphorylation-based Mechanism
Shohat G, et al.
Test, 276, 47460-47467 (2001)
A functional genetic screen identifies regions at the C-terminal tail and death-domain of death-associated protein kinase that are critical for its proapoptotic activity
Raveh T, et al.
Proceedings of the National Academy of Sciences of the USA, 97, 1572-1577 (2000)
Song Liu et al.
Frontiers in neuroscience, 15, 525615-525615 (2021-03-12)
Excitatory toxicity due to excessive glutamate release is considered the core pathophysiological mechanism of cerebral ischemia. It is primarily mediated by N-methyl-D-aspartate receptors (NMDARs) on neuronal membranes. Our previous studies have found that icaritin (ICT) exhibits neuroprotective effects against cerebral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service