Skip to Content
Merck
All Photos(1)

Key Documents

D4692

Sigma-Aldrich

Anti-DNMT1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DNA methyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

human, mouse

concentration

0.8-1.2 mg/mL

technique(s)

microarray: suitable
western blot: 0.5-1.0 μg/mL using nuclear extracts of 293T or HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNMT1(1786)

General description

In humans, the gene encodes a DNA (cytosine-5)-methyltransferase with 1616 amino acids. The N-terminal contains the regulatory domain, while the C-terminal region contains the catalytic domain. Two isoforms of DNMT1 have been isolated, DNMT1a and DNMT1b, the difference residing in 16 extra amino acids within the latter.

Immunogen

synthetic peptide corresponding to amino acids 72-78 of human DNMT1, conjugated to KLH via an N-terminal added cysteine residue. The sequence is conserved in mouse and is different from the rat sequence by one amino acid.

Application

Anti-DNMT1 antibody produced in rabbit is suitable for microarray and immunoblotting at a working concentration of 0.5-1.0μg/mL using nuclear extracts of 293T or HeLa cells. It was used at 1:1000 working dilution for immunoblot analysis in a study to analyze whether some miRNAs are aberrantly expressed and target DNMT1 in NiS (nickel sulfide)-transformed cells. It was used for immunoprecipitation of DNMT1 from nuclear lysates of colon cancer cells transfected with DNMT1 vectors in a study. It was used as a primary antibody at a working dilution of 1:1000 for western blot analysis of total protein extracted from left murine testis tissue exposed to low-dose-rate radiation in a study.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

DNMT1 protein is important for the maintenance of methylation, as well as for the novo methylation activities occurring in somatic cells of vertebrates. It establishes a repressive transcription complex at replication foci with histone deacetylase HDAAC4 and DMAP1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Ji Gong et al.
Journal of radiation research, 55(1), 54-60 (2013-09-13)
This study examined the effects of continuous low-dose-rate radiation exposure (3.49 mGy/h) of gamma rays on mice testicles. C57BL/6 mice were divided into sham and radiation groups (n = 8 each), and were exposed to either sham irradiation or 2
Weidong Ji et al.
Carcinogenesis, 34(2), 446-453 (2012-11-06)
Nickel (Ni) compounds are well-recognized human carcinogens, yet the molecular mechanisms by which they cause human cancer are still not well understood. MicroRNAs (miRNAs), which are small non-coding RNAs, are involved in diverse biological functions and carcinogenesis. In previous study
Anna G Manjón et al.
Cell reports, 42(10), 113124-113124 (2023-09-21)
Acquired drug resistance is a major problem in the treatment of cancer. hTERT-immortalized, untransformed RPE-1 cells can acquire resistance to Taxol by derepressing the ABCB1 gene, encoding for the multidrug transporter P-gP. Here, we investigate how the ABCB1 gene is
Haiqing Zhang et al.
Oncology reports, 40(3), 1803-1812 (2018-07-18)
Downregulation of microRNA‑152 (miR‑152) has been observed in various types of human malignancies, including Bladder cancer (BC). However, the role of miR‑152 in the development and progression of BC is still unclear. In our previous study, we identified a functional
Michael J Topper et al.
Cell, 171(6), 1284-1300 (2017-12-02)
Combining DNA-demethylating agents (DNA methyltransferase inhibitors [DNMTis]) with histone deacetylase inhibitors (HDACis) holds promise for enhancing cancer immune therapy. Herein, pharmacologic and isoform specificity of HDACis are investigated to guide their addition to a DNMTi, thus devising a new, low-dose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service