Skip to Content
Merck
All Photos(2)

Key Documents

D4668

Sigma-Aldrich

Dextranase from Penicillium sp.

lyophilized powder, 100-250 units/mg protein

Synonym(s):

1,6-α-D-Glucan 6-glucanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

100-250 units/mg protein

composition

Protein, ~25% Lowry

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dextranase from Penicillium sp. can be used to modify alternan. Dextranase from Penicillium sp. has also been used in a study that explored the possibility that the Dex protein might adopt a fold unfamiliar to any known structure.

Biochem/physiol Actions

The product is an endodextranase that hydrolyzes α-(1,6)-glucosidic linkages in dextran. Dextrans are undesirable compounds synthesized from sucrose by microbial contaminants during sugar production. They increase viscosity of the flow and decrease industrial recovery. Dextranase has been used for hydrolyzing dextran at sugar mills in order to improve efficiency of sugar production.

Unit Definition

One unit will liberate 1.0 μmole of isomaltose (measured as maltose) per min at pH 6.0 at 37 °C, using dextran as substrate.

Physical form

Partially purified, lyophilized powder

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Pons et al.
Proteins, 31(4), 345-354 (1998-06-17)
The DEX gene encodes an extracellular dextranase (EC 3.2.1.11); this enzyme hydrolyzes the alpha(1,6) glucosidic bond contained in dextran to release small isomaltosaccharides. Sequence analysis has revealed only one homologous sequence, CB-8 protein, from Arthrobacter sp., with 30% sequence identity.
Timothy D Leathers et al.
Biotechnology letters, 31(2), 289-293 (2008-10-22)
Alternan is a unique glucan with a backbone structure of alternating alpha-(1 --> 6) and alpha-(1 --> 3) linkages. Previously, we isolated strains of Penicillium sp. that modify native, high molecular weight alternan in a novel bioconversion process to a
Dextranase in sugar industry: A review
Efrain Rodriguez Jimenez
Sugar Tech, 11(2), 124-134 (2009)
Akira Yano et al.
Applied microbiology and biotechnology, 86(2), 615-623 (2009-11-11)
Mushrooms contain large quantities of alpha-glucans. Shiitake (Lentinula edodes), Japan's most popular edible mushroom, has been reported to contain about 6% (weight/dried weight) of alpha-(1,3)-glucan. This glucan is one of the major components of oral biofilm formed by the cariogenic
Hongbin Zhang et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 51(4), 495-503 (2011-07-30)
To obtain new fungi producing dextranase,we screened and identified a strain F1001 showing high dextranase activities. We provided a new strain with dextranase activity for producing clinical dextran. Morphological and ITS rDNA sequences homology analysis were performed to identify the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service