Skip to Content
Merck
All Photos(1)

Documents

D3168

Sigma-Aldrich

Duramycin from Streptoverticillium cinnamoneus

≥90.0%

Synonym(s):

Duramycin, Lancovutide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C89H125N23O25S3
CAS Number:
Molecular Weight:
2013.28
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85

Quality Level

Assay

≥90.0%

form

powder

solubility

0.1 M HCl: 10 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

General description

Chemical structure: tetracycline

Application

Duramycin is used to study chloride secretion by cardial, pancreatic and airway epithelial voltage-gated ion channels. It is used to study cystic fibrosis and as a novel phosphatidylethanolamine-binding molecular probe. It is used to stimulate sodium transport in cultured human colonic epithelia.

Biochem/physiol Actions

Duramycin is the smallest known polypeptide since it has only 19 amino acids. It has a defined 3-dimensional binding structure. Duramycin binds phosphatidylethanolamine (PtdE) at a 1:1 ratio with high affinity and exclusive specificity. It enhances chloride secretion in airway epithelium.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Zhao et al.
Nuclear medicine and biology, 39(7), 1006-1011 (2012-08-04)
(99m)Tc-Duramycin is a unique radiopharmaceutical that binds specifically to phosphatidylethanolamine (PE). The current effort is to develop a single-step kit formulation for the (99m)Tc labeling of HYNIC-Duramycin. A titration series of Tricine/TPPTS coligand systems were tested for an optimal formulation
Soichiro Matsunaga et al.
Langmuir : the ACS journal of surfaces and colloids, 25(14), 8200-8207 (2009-05-13)
We visualized nanometer-scale phospholipid particle fusion by scanning tunneling microscopy (STM) on an alkanethiol-modified gold substrate, induced by duramycin, a tetracyclic antibiotic peptide with 19 amino residues. Ultrasonic homogenization generated a suspension mainly consisting of minimal lipid particles (MLP) from
Ming Zhao et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 49(8), 1345-1352 (2008-07-18)
With only 19 amino acids, duramycin is the smallest known polypeptide that has a defined 3-dimensional binding structure. Duramycin binds phosphatidylethanolamine (PtdE) at a 1:1 ratio with high affinity and exclusive specificity. As an abundant binding target, PtdE is a
M M Cloutier et al.
The American journal of physiology, 259(3 Pt 1), C450-C454 (1990-09-01)
The effect of duramycin, a polypeptide antibiotic, on Cl- transport in canine tracheal epithelium mounted in Ussing chambers was studied. Over a narrow concentration range, duramycin increased short-circuit current (Isc) and net Cl- secretion and had no effect on mannitol
Igor Oliynyk et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 118(12), 982-990 (2010-11-26)
The lantibiotic duramycin (Moli1901, Lancovutide) has been suggested as a drug of choice in the treatment for cystic fibrosis (CF). It has been proposed that duramycin may stimulate chloride secretion through Ca²(+) -activated Cl⁻ channels (CaCC). We investigated whether duramycin

Articles

Ribosomally synthesized antimicrobial peptides are a promising focus in antibiotic research amidst bacterial resistance and emerging infectious diseases.

Ribosomally synthesized antimicrobial peptides are a promising focus in antibiotic research amidst bacterial resistance and emerging infectious diseases.

Ribosomally synthesized antimicrobial peptides are a promising focus in antibiotic research amidst bacterial resistance and emerging infectious diseases.

Ribosomally synthesized antimicrobial peptides are a promising focus in antibiotic research amidst bacterial resistance and emerging infectious diseases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service