Skip to Content
Merck
All Photos(1)

Key Documents

D3072

Sigma-Aldrich

Doxycycline Hydrochloride, Ready Made Solution

100 mg/mL in DMSO, ≥95.0% (HPLC)

Synonym(s):

α-6-Deoxy-5-hydroxytetracycline hydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride (1:1), (4S,4aR,5S,5aR,6R,12aS)-, Doxylin, Idocyklin, Vibramycin hyclate, Vibramycin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
MDL number:
UNSPSC Code:
51284028
NACRES:
NA.85

biological source

synthetic

Quality Level

sterility

0.22 μm filtered

Assay

≥95.0% (HPLC)

form

liquid

concentration

100 mg/mL in DMSO

solubility

DMSO: 100 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma
parasites

Mode of action

protein synthesis | interferes

storage temp.

−20°C

InChI

1S/C22H24N2O8.ClH/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-7,10,14-15,17,25-27,31-32H,23H2,1-3H3;1H/b21-13-;/t7-,10+,14+,15-,17-,22-;/m0./s1

InChI key

VLUQVUWDECWBTL-UQVCFKGQSA-N

General description

Doxycycline is a member of the tetracycline antibiotics group, and is commonly used to treat a variety of infections. It is a Broad spectrum antibiotic and bacteriostatic. Doxycycline shows antiprotozoal properties and is a potent inhibitor of MMPs (matrix metalloproteinases) in vivo.

Application

<ul>
<li><strong>AAPS PharmSciTech: </strong>Creation of a PVA/PCL/CS-based nanofibrous membrane incorporating doxycycline hydrochloride for guided tissue regeneration and controlled delivery in the management of periodontitis, evaluated in vivo in rats (Mirzaeei et al., 2024).</li>
<li><strong>European Journal of Pharmacology: </strong>Doxycycline hydrochloride′s role in inhibiting the progression of malignant rhabdoid tumor of the kidney by targeting MMP17 and MMP1 through the PI3K-Akt signaling pathway (Mi et al., 2024).</li>
</ul>

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youcai Tang et al.
PloS one, 13(8), e0200897-e0200897 (2018-08-02)
Alpha-1 Antitrypsin (α1AT) Deficiency is a genetic disease in which accumulation of α1AT mutant Z (α1ATZ) protein in the ER of hepatocytes causes chronic liver injury, liver fibrosis, and hepatocellular carcinoma. No effective medical therapy is currently available for the
Megan E Forrest et al.
PloS one, 15(2), e0228730-e0228730 (2020-02-14)
Messenger RNA (mRNA) degradation plays a critical role in regulating transcript levels in the cell and is a major control point for modulating gene expression. In yeast and other model organisms, codon identity is a powerful determinant of transcript stability
Adam W Whisnant et al.
Nature communications, 11(1), 2038-2038 (2020-04-29)
The predicted 80 open reading frames (ORFs) of herpes simplex virus 1 (HSV-1) have been intensively studied for decades. Here, we unravel the complete viral transcriptome and translatome during lytic infection with base-pair resolution by computational integration of multi-omics data.
Benedetta Assetta et al.
Journal of visualized experiments : JoVE, (165) (2020-11-24)
In Alzheimer's disease (AD) and other neurodegenerative disorders, oligodendroglial failure is a common early pathological feature, but how it contributes to disease development and progression, particularly in the gray matter of the brain, remains largely unknown. The dysfunction of oligodendrocyte
Steve Ramirez et al.
Science (New York, N.Y.), 341(6144), 387-391 (2013-07-28)
Memories can be unreliable. We created a false memory in mice by optogenetically manipulating memory engram-bearing cells in the hippocampus. Dentate gyrus (DG) or CA1 neurons activated by exposure to a particular context were labeled with channelrhodopsin-2. These neurons were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service