Skip to Content
Merck
All Photos(1)

Documents

D1947

Sigma-Aldrich

Diffinity RapidTip®

for PCR Purification

Synonym(s):

PCR clean up tip, PCR purification tip, PCR reaction clean-up tip

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106300
NACRES:
NA.52

Quality Level

form

solid

manufacturer/tradename

(Diffinity Genomics, Inc.)

capacity

50(µL)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Diffinity RapidTip effectively removes dNTPs, primers and primer dimers while providing greater than 90% recovery of pure DNA fragments from 100 bp to 10 kb. The functional pipette tip contains everything you need for PCR purification. The tip is filled with a proprietary adsorption technology that has a differential affinity for PCR components. The dispensed solution yields purified, high quality DNA ready for downstream applications such as DNA sequencing, SNP analysis and microarray printing.

Application

Diffinity RapidTip® has been used in PCR product purification.

Features and Benefits

  • Single step
  • Recovers 90% of high quality dsDNA
  • Optimized for 25 μL RXN

Legal Information

RapidTip is a registered trademark of Diffinity Genomics, Inc.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT RE 2 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa A Santry et al.
Virus research, 175(1), 30-44 (2013-04-16)
Maedi-visna virus (MVV) and caprine arthritis encephalitis virus (CAEV) are related members of a group of small ruminant lentiviruses (SRLVs) that infect sheep and goats. SRLVs are endemic in many countries, including Canada. However, very little is known about the
C N Roterman et al.
Proceedings. Biological sciences, 280(1764), 20130718-20130718 (2013-06-21)
The phylogeny of the superfamily Chirostyloidea (Decapoda: Anomura) has been poorly understood owing to limited taxon sampling and discordance between different genes. We present a nine-gene dataset across 15 chirostyloids, including all known yeti crabs (Kiwaidae), to improve the resolution
Myron T La Duc et al.
Applied and environmental microbiology, 78(16), 5912-5922 (2012-06-26)
Spacecraft hardware and assembly cleanroom surfaces (233 m(2) in total) were sampled, total genomic DNA was extracted, hypervariable regions of the 16S rRNA gene (bacteria and archaea) and ribosomal internal transcribed spacer (ITS) region (fungi) were subjected to 454 tag-encoded
Marion L East et al.
International journal for parasitology. Parasites and wildlife, 2, 257-265 (2014-02-18)
We provide the first genetic sequence data for a Dipylidium species from a wild carnivore plus an analysis of the effects of ecological, demographic, physiological and behavioural factors on Dipylidium sp. infection prevalence in a social carnivore, the spotted hyaena
H Kühlwein et al.
Journal of applied microbiology, 115(5), 1091-1106 (2013-07-31)
To assess the effects of dietary Saccharomyces cerevisiae β-(1,3)(1,6)-D-glucan supplementation (MacroGard(®)) on mirror carp (Cyprinus carpio L.) intestinal microbiota and ultrastructure of the enterocyte apical brush border. Carp were fed either a control diet or diets supplemented with 0.1, 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service