Skip to Content
Merck
All Photos(1)

Key Documents

D0301

Sigma-Aldrich

1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol

≥95% (TLC), liquid

Synonym(s):

1,2-Di(cis-9,12-octadecadienoyl)-3-hexadecanoyl-rac-glycerol, PLL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H98O6
CAS Number:
Molecular Weight:
855.36
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥95% (TLC)

form

liquid

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCCCC/C=C\C/C=C\CCCCC)OC(COC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C55H98O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h16-17,19-20,25-28,52H,4-15,18,21-24,29-51H2,1-3H3/b19-16-,20-17-,27-25-,28-26-

InChI key

LXAWUIOWWNQCQA-YBQWMRSQSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

Liquid at room temperature

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hirotaka Nakatsuji et al.
Scientific reports, 7(1), 4694-4694 (2017-07-07)
Light-inducible gene regulation has great potential for remote and noninvasive control of the fate and function of target cells. One method to achieve such control is delivery of heat shock protein (HSP) promoter-driven protein expression vectors and photothermal heaters into
Ju An Park et al.
Scientific reports, 7(1), 14610-14610 (2017-11-04)
Microfabrication methods have widely been used to control the local cellular environment on a micron scale. However, accurately mimicking the complexity of the in vivo tissue architecture while maintaining the freedom of form and design is still a challenge when
Sunita D Bansod et al.
IET nanobiotechnology, 13(6), 609-616 (2019-08-23)
Here, a rapid and easy transformation by electroporation technique for gene transfer in plants using cell penetrating amino nanocomplex (nanoplex) has been demonstrated in Nicotiana. Nanoplex was prepared using cell penetrating amino acids (CPAs) such as poly-L-lysine (PLL) and Argenine
Andrea J Mothe et al.
Scientific reports, 7(1), 10529-10529 (2017-09-07)
Traumatic spinal cord injury (SCI) causes a cascade of degenerative events including cell death, axonal damage, and the upregulation of inhibitory molecules which prevent regeneration and limit recovery. Repulsive guidance molecule A (RGMa) is a potent neurite growth inhibitor in
Satoshi Imai et al.
Scientific reports, 7(1), 5947-5947 (2017-07-22)
Impairment of peripheral neurons by anti-cancer agents, including taxanes and platinum derivatives, has been considered to be a major cause of chemotherapy-induced peripheral neuropathy (CIPN), however, the precise underlying mechanisms are not fully understood. Here, we examined the direct effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service