Skip to Content
Merck
All Photos(2)

Key Documents

CS0340

Sigma-Aldrich

Dihydrofolate Reductase Assay Kit

1 kit sufficient for 50-100 tests

Synonym(s):

DHFR Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

usage

 kit sufficient for 50-100 tests

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... DHFR(1719)

Application

The kit is used for the detection of DHFR activity and screening of DHFR inhibitors. The assay is based on the ability of DHFR to catalyze the reversible NADPH-dependent reduction of dihydrofolic acid to tetrahydrofolic acid. The reaction progress is followed by monitoring the decrease in absorbance at 340 nm.
Dihydrofolic acid+NADPH+H+ ↔ Tetrahydrofolic acid+NADP+

Biochem/physiol Actions

DHFR (dihydrofolate reductase) is an enzyme that catalyzes a reaction essential for the biosynthesis of nucleotidic bases of DNA. Blocking the enzyme causes cell death as a result of DNA synthesis inhibition. DHFR is an excellent target for anti-tumor drugs.

Features and Benefits

  • Quick and simple method.
  • The kit contains all the reagents required for a colorimetric assay of DHFR activity in cell lysates, tissue homogenates, or column fractions of purified enzyme.
  • The kit includes a purified enzyme for use as a positive control and screening of DHFR inhibitors.
  • The kit includes methotrexate (MTX), a prokaryotic and eukaryotic DHFR specific inhibitor, which exhibits anti-tumor activities.
  • The kit was tested on A431, NIH-3T3, and CHO cell lines, rat liver, kidney, brain, and skeletal muscle tissue extracts, and recombinant DHFR.

Kit Components Only

Product No.
Description

  • Assay Buffer 10x for DHFR 30 mL

  • Dihydrofolate Reductase (DHFR) human .1 U

  • Dihydrofolic acid (DHFR substrate) 3 x 10

  • Amethopterin (+)(methotrexate, MTX)
    (DHFR inhibitor) 2 x 10

  • NADPH (β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt) 25 mg

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amrinder Singh et al.
Scientific reports, 8(1), 3190-3190 (2018-02-18)
We report the first peptide based hDHFR inhibitors designed on the basis of structural analysis of dihydrofolate reductase (DHFR). A set of peptides were rationally designed and synthesized using solid phase peptide synthesis and characterized using nuclear magnetic resonance and
Bharath Srinivasan et al.
The FEBS journal, 282(10), 1922-1938 (2015-02-24)
Dihydrofolate reductase (DHFR) is a pivotal enzyme involved in the de novo pathway of purine synthesis, and hence, represents an attractive target to disrupt systems that require rapid DNA turnover. The enzyme acquires resistance to available drugs by various molecular
Marta Jorba et al.
Antibiotics (Basel, Switzerland), 10(6) (2021-07-03)
This work reports a detailed characterization of the antimicrobial profile of two trimethoprim-like molecules (compounds 1a and 1b) identified in previous studies. Both molecules displayed remarkable antimicrobial activity, particularly when combined with sulfamethoxazole. In disk diffusion assays on Petri dishes
Bermans J Iskandar et al.
The Journal of clinical investigation, 120(5), 1603-1616 (2010-04-29)
The folate pathway plays a crucial role in the regeneration and repair of the adult CNS after injury. Here, we have shown in rodents that such repair occurs at least in part through DNA methylation. In animals with combined spinal
Bharath Srinivasan et al.
European journal of medicinal chemistry, 103, 600-614 (2015-09-29)
Gram-negative bacteria are implicated in the causation of life-threatening hospital-acquired infections. They acquire rapid resistance to multiple drugs and available antibiotics. Hence, there is the need to discover new antibacterial agents with novel scaffolds. For the first time, this study

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service