Skip to Content
Merck

CRISPR

CRISPR/Cas9 Products and Services

Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.

Synonym(s):

Crispr RNA, Crispr/Cas System, CRISPR, CRISPRs, Cas9

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200



Takehito Kaneko et al.
Scientific reports, 4, 6382-6382 (2014-10-02)
Engineered endonucleases, such as zinc-finger nucleases (ZFNs), transcription activator-like effector nucleases (TALENs), and the clustered regularly interspaced short palindromic repeat (CRISPR)/CRISPR-associated (Cas) system, provide a powerful approach for genome editing in animals. However, the microinjection of endonucleases into embryos requires
Le Cong et al.
Science (New York, N.Y.), 339(6121), 819-823 (2013-01-05)
Functional elucidation of causal genetic variants and elements requires precise genome editing technologies. The type II prokaryotic CRISPR (clustered regularly interspaced short palindromic repeats)/Cas adaptive immune system has been shown to facilitate RNA-guided site-specific DNA cleavage. We engineered two different
Joshua R York et al.
Developmental biology, 453(2), 180-190 (2019-06-19)
A major challenge in vertebrate evolution is to identify the gene regulatory mechanisms that facilitated the origin of neural crest cells and placodes from ancestral precursors in invertebrates. Here, we show in lamprey, a primitively jawless vertebrate, that the transcription



Global Trade Item Number

SKUGTIN
CRISPR04061838113177
CRISPRPL-1EA04061833541401
HSOLIGOINT-1KT04061833826539
PCRISPR002-1KT04065266925241
PCRISPR001-1KT04065266925234
HKCRISPR-1EA04061833813959
CRISPR01-1SET04061833533802
CRISPR02-1SET04061833533819
PCRISPR003-1KT04065266925258
PCRISPR008-1EA04065266925302
PCRISPR007-1EA04065266925296
PCRISPR004-1EA04065266925265
CAS9D10AMRNA-1EA04061833531495
CAS9D10AP-1EA04061833531501
CRISPR06-1EA04061833533826
CRISPR07-1EA04061833533833
CRISPR08-1EA04061833533840
CRISPR12-1EA04061833541289
CRISPR11-1EA04061833541265