CMR0002
Recovery Medium for Cloning
for use with competent cells
Synonym(s):
Recovery Medium for Cloning
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41106200
NACRES:
NA.85
Recommended Products
grade
for molecular biology
sterility
sterile; autoclaved
shipped in
dry ice
storage temp.
−70°C
Features and Benefits
Use in the outgrowth stage of competent cell transformation to achieve 10-folder higher transformation efficiency than you would get with LB or SOC media.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service