Skip to Content
Merck
All Photos(1)

Key Documents

CLS92592

Corning® 1 x 8 Stripwell 96 well EIA plates

96 well plate, assembled 1 x 8 strips per 96 well plate, non-treated, non-sterile, 100/cs

Synonym(s):

1 x 8 strip plate, 8 well strip plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.15
material:
clear wells
size:
96 wells
sterility:
non-sterile
binding type:
non-treated surface
feature:
lid: no
plate format: 1 x 8 Stripwell

material

clear wells

sterility

non-sterile

feature

lid: no
plate format: 1 x 8 Stripwell

packaging

case of 100

manufacturer/tradename

Corning 92592

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

none

binding type

non-treated surface

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service