Skip to Content
Merck
All Photos(1)

Key Documents

CLS3748

Corning® Thermowell GOLD PCR 1 x 8 cap strips

Synonym(s):

1 x 8 cap strips, 8 well cap strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

polypropylene

sterility

non-sterile

feature

autoclavable
designed to fit on all Thermowell GOLD 1 x 8 well PCR strips and 96 well PCR microplates

packaging

pack of 125
pack of 1250

manufacturer/tradename

Corning 3748

technique(s)

PCR: suitable

color

assorted colors

suitability

suitable for (PCR applications)

Looking for similar products? Visit Product Comparison Guide

General description

  • Made of thin wall polypropylene and designed to fit on all Thermowell GOLD 1 x 8 well PCR strips and 96 well PCR microplates
  • Certified DNase- and RNase-free
  • Autoclavable at 121°C

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service