Key Documents
CLS2580
Corning® 1 x 8 Stripwell 96 well plates
high binding surface, without stripwell holder
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom clear polystyrene (96-well)
round wells
size:
96 wells
well volume:
360 μL
well working volume:
75-200 μL , for low volume plates volume is 50-125 μL
Pricing and availability is not currently available.
material
flat bottom clear polystyrene (96-well)
round wells
sterility
non-sterile
feature
lid: no
packaging
plate of 12 strips
manufacturer/tradename
Corning 2580
size
96 wells
well volume
360 μL
well working volume
75-200 μL , for low volume plates volume is 50-125 μL
color
clear
binding type
high binding surface
General description
The Costar 1 × 8 strips are pre-assembled in an "egg crate" style 96 well strip holder. Strips fit in holder only one way, eliminating mis-orientation. If covers are needed, the notched cover fits the fully assembled strip holder.
Legal Information
Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)