Skip to Content
Merck
All Photos(1)

Key Documents

CLS2481

Corning® 1 x 8 Stripwell 96 well plates

low volume, high binding surface, clear

Synonym(s):

1 x 8 strip plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear polystyrene plate
flat bottom clear polystyrene (96-well)
round clear wells
size:
96 wells
well volume:
190 μL
well working volume:
75-200 μL , for low volume plates volume is 50-125 μL
Pricing and availability is not currently available.

material

clear polystyrene plate
flat bottom clear polystyrene (96-well)
round clear wells

sterility

non-sterile

feature

lid: no
skirt

packaging

plate of 12 strips

manufacturer/tradename

Corning 2481

size

96 wells

well volume

190 μL

well working volume

75-200 μL , for low volume plates volume is 50-125 μL

color

clear

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The Costar 1 × 8 strips are pre-assembled in an "egg crate" style 96 well strip holder. Strips fit in holder only one way, eliminating mis-orientation. If covers are needed, the notched cover fits the fully assembled strip holder.

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service