Skip to Content
Merck

CLLS1069

MCF10A CELLS APC (-/-)

human female mammary glands (Source Disease: Fibrocystic Disease)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102
Biological source:
human female mammary glands (Source Disease: Fibrocystic Disease)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

MCF10A CELLS APC (-/-),

biological source

human female mammary glands (Source Disease: Fibrocystic Disease)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... APC(324)

General description

MCF10A CELLS APC (-/-) are pre-neoplastic mammary epithelial cells from a human adult female with a ZFN knock out modification.
This cell line corresponds to ATCC Catalog No. CRL-10317.
Adenomatous polyposis coli (APC) is a tumor suppressor gene. It is located on human chromosome 5q21.

Application

Cell freezing medium-DMSO 1X, Cat. No. C6164
Media renewal changes two to three times per week.
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°
The base medium for this cell line is Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F-12, Cat. No. 51448C. To make the complete growth medium, add the following components to the base medium: horse serum, Cat No. H1270, to a final concentration (v/v) of 5%; cholera toxin, Cat No. C8052, to a final concentration of 1 ng/mL; human insulin, Cat No. I9278, to a final concentration of 10 ug/mL; epidermal growth factor, Cat No. E9644, to a final concentration of 10 ng/mL; and hydrocortisone, Cat No. H6909, to a final concentration of 0.5 ug/mL.

Biochem/physiol Actions

Adenomatous polyposis coli (APC) plays an important role in cell division, adhesion and migration. Mutations in APC gene is associated with colorectal carcinomas.

Features and Benefits

These MCF10A cells are adherent with a doubling time of approx. 20 hours
Zinc finger nuclease (ZFN) knockout on chromosome 5q21

Analysis Note

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • MCF10A CONTROL CELLS 1 vialSDS

  • MCF10A CELLS APC (-/-) 1 vialSDS

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenomatous polyposis coli (APC) regulates miR17-92 cluster through β-catenin pathway in colorectal cancer.
Li Y, et al.
Oncogene, 35(35), 4558?4568-4558?4568 (2016)
Epigenetic regulation of APC in the molecular pathogenesis of gallbladder cancer.
Tekcham DS, et al.
The Indian Journal of Medical Research, 143(S1), S82?S90-S82?S90 (2016)

Related Content

Global Trade Item Number

SKUGTIN
CLLS1069-1SET04061833532270

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service