CD1
Cell dissociation sieve - tissue grinder kit
tissue grinder homogenizer kit, stainless steel, autoclavable, 1/kit
Synonym(s):
tissue grinder homogenizer, tissue homogenizer, tissue pulverizer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
packaging
kit of 1
Components
Tissue Grinder Homogenizer Kit
Components include:
Kit contains one 85 mL cup, 5 each of 40, 50 and 60 mesh screens, 5 screen replacement keys and 2 glass pestles.
Components include:
Kit contains one 85 mL cup, 5 each of 40, 50 and 60 mesh screens, 5 screen replacement keys and 2 glass pestles.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 13(8), e0201377-e0201377 (2018-08-03)
In recent years, the traditional treatments for thrombotic diseases, heparin and warfarin, are increasingly being replaced by novel oral anticoagulants offering convenient dosing regimens, more predictable anticoagulant responses, and less frequent monitoring. However, these drugs can be contraindicated for some
Development (Cambridge, England), 144(9), 1698-1711 (2017-03-31)
Regulated retinal ganglion cell (RGC) differentiation and axonal guidance is required for a functional visual system. Homeodomain and basic helix-loop-helix transcription factors are required for retinogenesis, as well as patterning, differentiation and maintenance of specific retinal cell types. We hypothesized
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(41), 9819-9827 (2017-09-08)
It has been reported consistently that many female chronic pain sufferers have an attenuation of symptoms during pregnancy. Rats display increased pain tolerance during pregnancy due to an increase in opioid receptors in the spinal cord. Past studies did not
Pain, 160(2), 358-366 (2018-10-20)
The processing of pain in the central nervous system is now known to have an important immune component, including T cells of the adaptive immune system. T cells have been shown to release endogenous opioids, and although it is well
Nature communications, 8(1), 2093-2093 (2017-12-14)
Teichoic acid (TA), a crucial cell wall constituent of the pathobiont Streptococcus pneumoniae, is bound to peptidoglycan (wall teichoic acid, WTA) or to membrane glycolipids (lipoteichoic acid, LTA). Both TA polymers share a common precursor synthesis pathway, but differ in
Related Content
Particle size conversion table from our Lab Basics Technical Library
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service