SHP-77
98110201, human lung, Not specified
Synonym(s):
Human SHP-77, SHP-77 Cell Line
About This Item
Recommended Products
Product Name
SHP-77, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 98110201, human lung, Not specified, 98110201
biological source
human lung
packaging
tube of 5 μg 98110201-DNA-5UG
pkg of vial of cells 98110201-1VL
growth mode
Adherent/aggregates in suspension
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Application
DNA Profile
CSF1PO: 10,11
D13S317: 8
D16S539: 11
D5S818: 12
D7S820: 11
THO1: 7
TPOX: 9,11
vWA: 16
Culture Medium
Subculture Routine
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service