Skip to Content
Merck
All Photos(1)

Documents

FGC4

94101904, rat skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

FGC4, 94101904

biological source

rat skin

growth mode

Adherent

karyotype

Modal no. 52

morphology

Epithelial

products

Albumin and ß-fibrogen

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat hepatoma [alb-gpt plasmid]

Cell Line Description

A subclone derived from Fao which is deficient in HGPRT (resistant to 5μg/ml 6-thioguanine and also resistant to 3mM ouabain). FGC4 contains the alb-gpt plasmid which is the bacterial gpt gene under control of the rat albumin promoter (400 bp). Produces albumin and ß-fibrinogen.GMO1

Application

Gene expression studies.

Culture Medium

Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures (70%) 1:100 i.e. seeding at 2x1000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Media change three times a week.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service