Skip to Content
Merck
All Photos(1)

Documents

HeLa Cell Line human

93021013, human cervix, Epithelial

Synonym(s):

HeLa Cells, HeLa-CCL2 Cells, Henrietta Lacks Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HeLa Cell Line human, epitheloid cervix carcinoma

biological source

human cervix

growth mode

Adherent

karyotype

aneuploidy

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix epitheloid carcinoma

Cell Line Description

Derived from a cervical carcinoma from a 31 year old female. This was the first aneuploid line derived from human tissue maintained in continuous cell culture. Susceptible to Poliovirus type I and adenovirus type 3. Identified as a contaminant in many other cell lines. The cells should be handled under laboratory containment level 2. Ethnicity: Black.

Application

HeLa cell line has been used to study whether Zika virus (ZIKV) infection alters centrosome number and spindle positioning in neural progenitor cells. It has also been used to evaluate the cytotoxicity of extracts, fruits and jams from Sorbus sps.
Virus studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 12,14
D16S539: 9,10
D5S818: 11,12
D7S820: 8,12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino AcidsNEAA + 10% Foetal Bovine Serum FBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1.3x10,000 cells/cm2 using 0.25% Trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.

Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.

Learn how to quantify spheroid formation, roundness, and circularity. Discover Millicell® Ultra-low Attachment plates, an ideal 3D cell culture model for uniform spheroid formation.

Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.

See All

Related Content

Millicell® Ultra-low Attachment plates facilitate 3D cell culture with detailed application information for cell seeding to analysis.

Millicell® Ultra-low Attachment plates facilitate 3D cell culture with detailed application information for cell seeding to analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service