Skip to Content
Merck
All Photos(1)

Key Documents

H9

85050301, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

H9, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 85050301

biological source

human blood

packaging

tube of 5 μg 85050301-DNA-5UG
pkg of vial of cells 85050301-1VL

growth mode

Suspension

karyotype

Modal no. 69

morphology

Lymphoblast

products

CD3, CD4

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T cell

Cell Line Description

Clonal derivative of the T-cell line HUT 78 derived from the blood of a patient with Sezary syndrome. The clone was selected due to its suitability for the isolation and continuous high level production and HIV variants with patients within AIDS and pre-AIDS. Examination with DNA probes indicate identical HLA typing and DNA fingerprints as HUT-78 .

Application

Detection, isolation and propagation of HIV-1

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11
D13S317: 8,12
D16S539: 11,12
D5S818: 11
D7S820: 8,11
THO1: 8,9
TPOX: 8,9
vWA: 14,15

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service