Skip to Content
Merck
All Photos(1)

Documents

C9992

Sigma-Aldrich

Anti-Caveolin-2 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution, suitable for western blot: 1 μg/mL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~20 kDa

species reactivity

pig (predicted), mouse, rat, canine, human (predicted)

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using whole extract of mouse 3T3-L1 adipocytes or rat kidney NRK cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAV2(858)
mouse ... Cav2(12390)
rat ... Cav2(363425)

General description

Caveolin-2 is an isoform of caveolin, an integral transmembrane protein of the caveolae membranes. It has a molar mass of 20-22 kDa and is a major coat protein found in caveolae of most cell types. It exists as two isoforms α and β due to alternative splicing of the respective mRNAs. Caveolin-2α corresponds to the full-length protein (20 kDa, 162 amino acids), whereas caveolin-2β isoform lacks a 13 amino acid sequence at its N-terminus.

Immunogen

synthetic peptide corresponding to amino acids 1-20 located at the N-terminus of mouse caveolin-2, conjugated to KLH. This sequece is identical in rat caveolin-2, highly conserved (90% identity) in human, dog, and pig caveolin-2, and is not found in caveolin-1 and caveolin-3.

Biochem/physiol Actions

Caveolin-2 colocalizes and forms a hetero-oligomeric complex with caveolin-1 in vivo. Caveolins family members function as scaffolding proteins to organize and concentrate specific lipids such as cholesterol and glycosylphosphatidylinositol (GPI) and lipid modified signaling molecules within caveolae membranes. Caveolin-2 is phosphorylated on Ser23 and Ser36 and modulates caveolin-1-dependent caveola formation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romina Paula Bulacio et al.
Journal of cellular biochemistry, 120(4), 4966-4974 (2018-10-01)
Caveolin-2 (Cav-2) is expressed in a variety of cell tissue, and it has also been found in renal tissue. The expression of Cav-2 in proximal tubules is still unclear. The aim of this study was to carry out a complete
P E Scherer et al.
The Journal of biological chemistry, 272(46), 29337-29346 (1997-11-20)
Caveolae are microdomains of the plasma membrane that have been implicated in organizing and compartmentalizing signal transducing molecules. Caveolin, a 21-24-kDa integral membrane protein, is a principal structural component of caveolae membrane in vivo. Recently, we and other laboratories have
R G Anderson
Annual review of biochemistry, 67, 199-225 (1998-10-06)
The cell biology of caveolae is a rapidly growing area of biomedical research. Caveolae are known primarily for their ability to transport molecules across endothelial cells, but modern cellular techniques have dramatically extended our view of caveolae. They form a
S Li et al.
The Journal of biological chemistry, 271(1), 568-573 (1996-01-05)
Caveolin, a 22-24-kDa integral membrane protein, is a principal component of caveolar membranes in vivo. Caveolin has been proposed to function as a scaffolding protein to organize and concentrate signaling molecules within caveolae. Because of its unusual membrane topology, both
M Sargiacomo et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(20), 9407-9411 (1995-09-26)
A 22-kDa protein, caveolin, is localized to the cytoplasmic surface of plasma membrane specializations called caveolae. We have proposed that caveolin may function as a scaffolding protein to organize and concentrate signaling molecules within caveolae. Here, we show that caveolin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service