Skip to Content
Merck
All Photos(1)

Key Documents

C8554

Sigma-Aldrich

Chorionic gonadotropin human

lyophilized powder, suitable for mouse embryo cell culture

Synonym(s):

Choriogonin, HCG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
biological source:
human urine
form:
lyophilized powder

biological source

human urine

Quality Level

sterility

aseptically processed

form

lyophilized powder

potency

≥5,000 I.U. per mg

mol wt

37.9 kDa
apparent mol wt 37.9 kDa by SDS-PAGE

drug control

Home Office Schedule 4.2

technique(s)

cell culture | embryo: suitable

impurities

HBV,HCV and HIV, tested negative

solubility

H2O: 1 mL/vial
saline: 1 mL/vial

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CGA(1081)

Looking for similar products? Visit Product Comparison Guide

General description

Glycoprotein hormone consisting of a 92-amino acid α-chain which is identical to that of LH, FSH, and TSH; and a distinct 149-amino acid β-chain. Produced by chorionic tissue and responsible for maintaining the corpus luteum during early pregnancy. Also produced by trophoblast cell neoplasms, which are marked by elevated free hCG β-chains and hyperglycosylated hCG.
Note: Actual molecular weight will vary from the theoretical depending upon the degree of glycosylation. Theoretical MW is 37.9 kDa based on native form which contains 2 subunits: α = 14.9 kDa (~10.2 kDa for the polypeptide and ~4.7 kDa for the carbohydrate), and β = 23.0 kDa (~16.0 kDa for the polypeptide and ~7.0 kDa for the carbohydrate). See J. Biol. Chem. 248, 6796-6827 (1973).

Application

Recommended for superovulation induction in mice.

Biochem/physiol Actions

Human chorionic gondadotropin (hCG) may be used as an alternative to PMSG to induce ovulation in laboratory animals. When hCG was injected into laboratory animals via intravenous (i.v.), intraperitoneal (i.p.), or subcutaneous (s.c.) methodstet, the effective dose values for 50% of the animals (ED50) per animal and per body weight (kg) in parenthesis were as follows:
Mice: i.v. 0.2 (7.7) I.U.; i.p. 0.3 (11.5) I.U.; s.c. 0.7 (26.9) I.U.
Syrian hamsters: i.v. 1.0 (9.5) I.U.; i.p. 1.8 (17.1) I.U.; s.c. 2.6 (24.8) I.U.
Rats: i.v. 1.3 (4.6) I.U.; i.p. 3.5 (12.3) I.U.; s.c.7.5 (26.3) I.U.

Using PMSG, the ED50 values per animal and per body weight (kg) in parenthesis were as follows:
Mice: i.v. 0.8 (30.8) I.U.; i.p. 2.0 (76.9) I.U.; s.c. 2.8 (107.7) I.U
Syrian hamsters: i.v., 3.6 (34.3) I.U.; i. p., 8.0 (76.2) I.U.; s.c., 13.2 (125.7) I.U.
Rats: i.v., 6.0 (76.8) I.U.; i p., 20.8 (73.0) I.U.; s.c., 76.8 (269.5) I.U.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Genetic-engineered models of skeletal diseases. II. Targeting mutations into transgenic mice chondrocytes.
S Garofalo et al.
Methods in molecular biology (Clifton, N.J.), 137, 491-498 (2000-08-19)
Zheng Zhang et al.
The Journal of reproduction and development, 55(1), 94-97 (2008-11-22)
Embryos transfer is one of the most critical steps for generating genetically modified mice. Because of the limitations of the current transfer equipment and techniques, such as discharge of freshly transferred embryos and the necessity of sophisticated transfer skills, the
Geetika Bassi et al.
iScience, 25(4), 104165-104165 (2022-04-19)
Mitochondria are essential for steroidogenesis. In steroidogenic cells, the initiation of steroidogenesis from cholesterol occurs on the matrix side of the inner mitochondrial membrane by the enzyme P450scc. This requires cholesterol import from the cytoplasm through the outer mitochondrial membrane
Manyu Li et al.
Biology of reproduction, 79(6), 1169-1175 (2008-08-22)
Adrenomedullin (AM) is a multifunctional peptide vasodilator that signals through a G-protein-coupled receptor when the receptor, called calcitonin receptor-like receptor (CL), is associated with a receptor activity-modifying protein 2 (RAMP2). We demonstrated previously that haploinsufficieny for each of these genes
Hui Yang et al.
Nature protocols, 9(8), 1956-1968 (2014-07-25)
Mice with specific gene modifications are valuable tools for studying development and disease. Traditional gene targeting in mice using embryonic stem (ES) cells, although suitable for generating sophisticated genetic modifications in endogenous genes, is complex and time-consuming. We have recently

Articles

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service