Skip to Content
Merck
All Photos(3)

Documents

C8241

Sigma-Aldrich

Chitinase from Trichoderma viride

lyophilized powder, ≥600 units/g solid

Synonym(s):

N-acetyl-β-glucosaminidase and chitodextrinase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥600 units/g solid

mol wt

30 kDa

solubility

0.05 M phosphate buffer, pH 6.0: soluble 0.90-1.10 mg/mL, faintly hazy to hazy (with particles)

shipped in

wet ice

storage temp.

−20°C

General description

Chitinase is an extracellular complex of enzymes that degrade chitin. Chitin is a cell wall component of Fungi and exoskeketal essentials of different organisms which reshape their own chitin or digest/dissolve the chitin of other organisms (insects, fungi, yeast, and algae, and in the internal structures of other vertebrates) . Chitinases have been detected in many microorganisms and in plants. In fungi, chitinases assist in morphogenesis, to break down the inherent chitin content of fungal cell walls. Plant chitinases help in resistance to fungal attack and counteracting fungal growth, by targeting those same fungal cell walls. In bacteria, bacterial chitinases assist in utilizing chitin as a carbon source and as an energy source.Streptomyces griseus produces multiple chitinases of different molecular masses after growth induction with chitin as the carbon source.

The enzymatic hydrolysis of chitin to N-acetyl-D-glucosamine involves two consecutive enzyme reactions:
  • The first reaction, chitodextrinase-chitinase, is a poly(β-(1→4)-[2-acetamido-2-deoxy-D-glucoside])- glycanohydrolase, which removes chitobiose units from chitin.
  • The second activity is N-acetyl-glucosaminidasechitobiase, which cleaves the disaccharide to its monomer subunits, N-acetyl-D-glucosamine.

Application

Agriculture fields: control pathogens.
Human health care: Asthma.
Pharma: preparation of chitooligosaccharides and N-acetyl D glucosamine,
Preparation of single-cell protein
Isolation of protoplasts from fungi and yeast
Control of pathogenic fungi
Treatment of chitinous waste, mosquito control and morphogenesis

Biochem/physiol Actions

Chitinase is a 30 kDa (approx.) extracellular enzyme complex that degrades chitin. Chitin is degraded to N-acetyl-D-glucosamine in 2 enzymatic reactions. Firstly, chitobiose units are removed from chitin by chitodextrinase-chitinase, a poly(1,4-β-[2-acetamido-2-deoxy-D-glucoside])-glycanohydrolase. The second reaction involves N-acetyl-glucosaminidase-chitobiase, which cleaves the disaccharide to its monomer subunits of N-acetyl-D-glucosamine. The enzyme may be classified into endo- and exochitinase. The endochitinase activity involves random cleavage at internal points in the chitin chain. The exochitinase activity consists of a progressive action which starts at the non-reducing end of chitin and releases chitobiose or N-acetyl-glucosamine units. The chitinolytic enzymes from T. viride are a mixture of extracellular chitinolytic enzymes, which exhibit exo- and endochitinase activities. The major activity was found to be that of N-acetyl-β-glucosaminidase.
Chitinase serves as a biopesticide against several fungi and insects. This hydrolytic enzyme is capable of cleaving the glycosidic bonds in chitin.

Features and Benefits

Chitinase is an extracellular complex of enzymes that degrade chitin. It is a lytic enzyme suitable for fungal cell walls lysis.

Unit Definition

One unit will liberate 1.0 mg of N-acetyl-D-glucosamine from chitin per hour at pH 6.0 at 25 °C in a 2 hour assay.
One new 1 hour unit = approx. 50 old 48 hour units.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A gene-targeting system for Pleurotus ostreatus: demonstrating the predominance of versatile-peroxidase (mnp4) by gene replacement
Salame TM, et al.
Applied and Environmental Microbiology, AEM-01234 (2012)
Sonia Żółtowska-Aksamitowska et al.
International journal of biological macromolecules, 112, 1021-1028 (2018-02-17)
Among marine demosponges (Porifera: Demospongiae), only representatives of the order Verongiida have been recognized to synthetize both biologically active substances as well as scaffolds-like fibrous skeletons made of structural aminopolysaccharide chitin. The unique 3D architecture of such scaffolds open perspectives
Production of microbial chitinases-a revisit.
PA Felse, T Panda
Bioprocess and Biosystems Engineering, 23, 127-134 (2000)
Functional Microbial Diversity: Functional Genomics and Metagenomics Using MAPLE
Microbial Diversity in the Genomic Era, 427-449 (2019)
Chitinases
Enzymes in Human and Animal Nutrition, 361-378 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service