Skip to Content
Merck
All Photos(1)

Key Documents

C7744

Sigma-Aldrich

Combretastatin A4

≥98% (HPLC), powder

Synonym(s):

1-(3,4,5-Trimethoxyphenyl)-2-(3′-hydroxy-4′-methoxyphenyl) ethane 3,4,5-trimethoxy-3′-hydroxy-4′-methoxystilbene, 2-Methoxy-5-[(1Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol, CA4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O5
CAS Number:
Molecular Weight:
316.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

COc1ccc(\C=C/c2cc(OC)c(OC)c(OC)c2)cc1O

InChI

1S/C18H20O5/c1-20-15-8-7-12(9-14(15)19)5-6-13-10-16(21-2)18(23-4)17(11-13)22-3/h5-11,19H,1-4H3/b6-5-

InChI key

HVXBOLULGPECHP-WAYWQWQTSA-N

General description

Combretastatin A4 belong to the class of colchicinoids compounds.

Application

Combretastatin A4 has been used:
  • as an anti-tubulin agent to determine the effects of isocitrate dehydrogenases (IDH1 and IDH2) proteins in G2/M phase
  • to evaluate the anti-proliferative and pro-apoptotic properties of biphenyl CA4 derivatives in both 2D and 3D cancerous and non-cancerous cell models
  • as a microtubule inhibitor to study its effects on motility of Ascaris suum L3 larvae

Biochem/physiol Actions

Combretastatin A4 is a potent microtubule targeting agent (MTA).
Combretastatin A4 is a vascular disrupting agent (VDA) that targets tumor vasculature to inhibit angiogenesis.
Combretastatin A4 is a vascular disrupting agent (VDA) that targets tumor vasculature to inhibit angiogenesis. It inhibits tubulin polymerization at the colchicine-binding site of beta-tubulin. It has antitumor activity by inhibiting AKT function in human gastric cells. The inhibited AKT activation causes decreased cell proliferation, cell cycle arrest, and reduced in vitro migration/invasiveness and in vivo metastatic ability. Combretastatin A4 is a natural stilbenoid phenol.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Thomas Nielsen et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(23), 6469-6477 (2012-10-17)
Combretastatin A-4 disodium phosphate (CA4P) is a promising vascular disrupting agent (VDA) in clinical trials. As CA4P acts on dividing endothelial cells, we hypothesize that CA4P affects vessels of certain sizes. The aim of this study was to evaluate the
Lisa M Greene et al.
Biochemical pharmacology, 84(5), 612-624 (2012-06-19)
Recent clinical data demonstrated that the vascular targeting agent Combretastatin-A4 phosphate (CA-4P) prolonged survival of patients with advanced anaplastic thyroid cancer without any adverse side effects. However, as a single agent CA-4 failed to reduce tumour growth in the murine
Eric Y Huang et al.
Journal of cellular and molecular medicine, 22(7), 3661-3670 (2018-04-24)
Single nucleotide polymorphisms (SNPs) within the regulatory elements of a gene can alter gene expression, making these SNPs of prime importance for candidate gene association studies. We aimed to determine whether such regulatory variants are associated with clinical outcomes in
Dong Liang et al.
Acta pharmaceutica Sinica. B, 12(5), 2429-2442 (2022-06-02)
Head and neck squamous cell carcinoma (HNSCC) is one of the most common human cancers; however, its outcome of pharmacotherapy is always very limited. Herein, we performed a batch query in the connectivity map (cMap) based on bioinformatics, queried out
Structurally simplified biphenyl combretastatin A4 derivatives retain in vitro anti-cancer activity dependent on mitotic arrest
Tarade D, et al.
Testing, 12(3) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service