Skip to Content
Merck
All Photos(1)

Key Documents

C7117

Sigma-Aldrich

Anti-Cullin-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cul1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
9
technique(s):
western blot: 0.5-1 μg/mL using whole cell extract of human colon carcinoma HCT-116 cell line and whole cell extract of mouse fibroblasts NIH3T3 cell line

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

species reactivity

human, mouse

technique(s)

western blot: 0.5-1 μg/mL using whole cell extract of human colon carcinoma HCT-116 cell line and whole cell extract of mouse fibroblasts NIH3T3 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL1(8454)
mouse ... Cul1(26965)

General description

Cullin-1 (Cul1) is encoded by the gene mapped to human chromosome 7q36.1. It is a 85 kDa protein that belongs to the family of cullin protein family containing distinct group of mediators (at least eight cullin members Cul1-8).

Immunogen

synthetic peptide located at the C-terminus of human cullin-1 (amino acids 759-776). This sequence is identical in mouse cullin-1 and highly conserved (83% identity) in Drosophila cullin-1.

Application

Anti-Cullin-1 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Cullin 1(CUL1) is a scaffold protein of the ubiquitin E3 ligase Skp1/Cullin1/Rbx1/F-box protein complex, which ubiquitinates a broad range of proteins that are involved in cell-cycle progression, signal transduction and transcription. Cul1 plays an important role in protein degradation and protein ubiquitination. It regulates breast cancer cell proliferation, migration and invasion. Overexpression of Cul1 leads to breast cancer progression. CUL1 promotes human trophoblast cell invasion and dysregulation of its expression may be associated with pre-eclampsia (PE). It may have some role in rheumatoid arthritis (RA) and its response to methotrexate. Cullin1 may act as an important marker for human gastric cancer lymph node metastasis and prognosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cullin-1 regulates MG63 cell proliferation and metastasis and is a novel prognostic marker of osteosarcoma
Cheng Q and Yin G
The International Journal of Biological Markers, 32(2), 202-209 (2017)
J Bai et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 24(8), 2016-2022 (2013-04-18)
To investigate the role of Cullin1 (Cul1) in the development of breast cancer, we examined the expression of Cul1 in breast cancer tissues and analyzed the correlation between Cul1 expression and clinicopathologic variables and patients survival. We evaluated the Cul1
Cullin 1 functions as a centrosomal suppressor of centriole multiplication by regulating polo-like kinase 4 protein levels
Korzeniewski N, et al.
Cancer research, 69(16), 6668-6675 (2009)
Jin Bai et al.
Human pathology, 42(3), 375-383 (2010-12-31)
Cullin1 (Cul1) is a scaffold protein of the ubiquitin E3 ligase Skp1/Cullin1/Rbx1/F-box protein complex, which ubiquitinates a broad range of proteins involved in cell-cycle progression, signal transduction, and transcription. To investigate the role of Cullin1 in the development of gastric
Sapna Negi et al.
Pharmacogenetics and genomics, 21(9), 590-593 (2011-07-09)
Aberrations in ubiquitin pathway have been implicated in many diseases and drug response. In a previous study on rheumatoid arthritis (RA) in Japanese population, significant association of Cullin1 gene (CUL1), an ubiquitin E3 ligase, was observed. CUL1 also mediates degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service