Skip to Content
Merck

Skip To

C6891

5-Chloro-2′-deoxyuridine

≥98% (HPLC), powder

Synonym(s):

CldU

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H11ClN2O5
CAS Number:
Molecular Weight:
262.65
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
MDL number:
eCl@ss:
32119108
Assay:
≥98% (HPLC)
Form:
powder
Solubility:
1 M NH4OH: 20 mg/mL, clear, colorless to faintly yellow
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5-Chloro-2′-deoxyuridine, thymidine analog

Quality Segment

assay

≥98% (HPLC)

form

powder

solubility

1 M NH4OH: 20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(Cl)C(=O)NC2=O

InChI

1S/C9H11ClN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

NJCXGFKPQSFZIB-RRKCRQDMSA-N

Application

5-Chloro-2′-deoxyuridine has been used to:
  • study cell cycle dynamics by immunohistochemical analysis and double S-phase labeling using animal models
  • analyze cell cycle length using an animal model
  • label HEK293T cells for molecular combing assay

Biochem/physiol Actions

DNA labeled with 5-chloro-2′-deoxyuridine (CldU) serves as an effective tool to analyze and quantify DNA replication, repair, and recombination. CldU is a potent mutagen, clastogen, and toxicant. It is used as a thymidine analog and is found to alter the dNTP pools and might lead to cell-cycle arrest. CldU produces sister-chromatid exchange but has less response to ionizing radiation compared to other thymine analogs. 5-Chloro-2′-deoxyuridine (CldU) is used to study the miscoding potential of hypochlorous acid damage to DNA and DNA precursors. When used with antibody based immunofluorescent imaging, 5-Chloro-2′-deoxyuridine incorporation may be used in protocols to identify sites of DNA replication. CldU may be used as a labeling substrate in conjunction with other halogenated uridine labeling substrates such as iododeoxyuridine (IdU).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D5412F0503343333
description

thymidine analog

description

≥98.5%

description

thymidylate synthase inhibitor

description

Experimental anticancer agent shown to have activity against a variety of malignant neoplasms, including mouse mammary tumors and colorectal carcinomas.

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

100

assay

≥98% (HPLC)

assay

≥98.5%

assay

≥99% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

solid

storage temp.

−20°C

storage temp.

-

storage temp.

room temp

storage temp.

2-8°C

solubility

1 M NH4OH: 20 mg/mL, clear, colorless to faintly yellow

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

water: 1 mg/mL, aqueous buffer: 1 mg/mL


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters